Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 61676879-61677003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL3
Entrez Gene ID
23284
Gene Name
adhesion G protein-coupled receptor L3
Gene Aliases
CIRL3, CL3, LEC3, LPHN3
UniGene
Hs.28391
Gene Chromosome Location
Chr.4: 61201256-62078335 on Build GRCh38
Transcripts
NM_001322246.1 NM_001322402.1 NM_015236.5 XM_011531788.2 XM_011531791.2 XM_017007929.1 XM_017007930.1 XM_017007931.1 XM_017007932.1 XM_017007933.1 XM_017007934.1 XM_017007935.1 XM_017007936.1 XM_017007937.1 XM_017007938.1 XM_017007939.1 XM_017007940.1 XM_017007941.1 XM_017007942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3604781 c.374C>T p.P125L Chr.4: 61676930-61676930 on Build GRCh38
6702503 c.357G>A p.V119V Chr.4: 61676913-61676913 on Build GRCh38
3302725 c.339C>A p.T113T Chr.4: 61676895-61676895 on Build GRCh38
387436 c.352G>A p.E118K Chr.4: 61676908-61676908 on Build GRCh38
4698515 c.350T>C p.L117P Chr.4: 61676906-61676906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376741663 Chr.4: 61676972-61676972 on Build GRCh38
rs182470778 Chr.4: 61676973-61676973 on Build GRCh38
rs373072697 Chr.4: 61676968-61676968 on Build GRCh38
rs1000308652 Chr.4: 61676965-61676965 on Build GRCh38
rs200275777 Chr.4: 61676958-61676958 on Build GRCh38
rs528117329 Chr.4: 61676915-61676915 on Build GRCh38
rs777412679 Chr.4: 61676950-61676950 on Build GRCh38
rs760041423 Chr.4: 61676954-61676954 on Build GRCh38
rs764402674 Chr.4: 61676965-61676966 on Build GRCh38
rs1041753886 Chr.4: 61676913-61676913 on Build GRCh38
rs764851224 Chr.4: 61676907-61676907 on Build GRCh38
rs951672990 Chr.4: 61676974-61676974 on Build GRCh38
rs751416704 Chr.4: 61676951-61676951 on Build GRCh38
rs748360580 Chr.4: 61676978-61676978 on Build GRCh38
rs758137316 Chr.4: 61676945-61676945 on Build GRCh38

View Full Product Details