Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4988726-4988981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INCA1
Entrez Gene ID
388324
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Gene Aliases
HSD45
UniGene
Hs.658926
Gene Chromosome Location
Chr.17: 4988130-4997933 on Build GRCh38
Transcripts
NM_001167985.1 NM_001167986.1 NM_001167987.1 NM_213726.2 XM_005256627.3 XM_005256628.4 XM_005256629.1 XM_006721517.1 XM_006721518.2 XM_006721519.3 XM_011523832.2 XM_011523834.2

Gene Symbol
LOC101927979
Entrez Gene ID
101927979
Gene Name
translation initiation factor IF-2
Gene Aliases
-
UniGene
Hs.673360
Gene Chromosome Location
Chr.17: 4986991-4990556 on Build GRCh38
Transcripts
NM_001167985.1 NM_001167986.1 NM_001167987.1 NM_213726.2 XM_005256627.3 XM_005256628.4 XM_005256629.1 XM_006721517.1 XM_006721518.2 XM_006721519.3 XM_011523832.2 XM_011523834.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546382 c.399G>A p.L133L Chr.17: 4988941-4988941 on Build GRCh38
4546383 c.354G>A p.L118L Chr.17: 4988941-4988941 on Build GRCh38
3766131 c.459G>C p.E153D Chr.17: 4988881-4988881 on Build GRCh38
4692458 c.489C>T p.D163D Chr.17: 4988806-4988806 on Build GRCh38
3766132 c.414G>C p.E138D Chr.17: 4988881-4988881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777787783 Chr.17: 4988888-4988888 on Build GRCh38
rs775202721 Chr.17: 4988804-4988804 on Build GRCh38
rs753534043 Chr.17: 4988895-4988895 on Build GRCh38
rs139444274 Chr.17: 4988957-4988957 on Build GRCh38
rs763211332 Chr.17: 4988858-4988858 on Build GRCh38
rs777365069 Chr.17: 4988752-4988752 on Build GRCh38
rs778661739 Chr.17: 4988778-4988778 on Build GRCh38
rs769214786 Chr.17: 4988807-4988807 on Build GRCh38
rs372170640 Chr.17: 4988861-4988861 on Build GRCh38
rs778618489 Chr.17: 4988926-4988926 on Build GRCh38
rs749123460 Chr.17: 4988755-4988755 on Build GRCh38
rs759371769 Chr.17: 4988825-4988825 on Build GRCh38
rs771689812 Chr.17: 4988794-4988794 on Build GRCh38
rs758195844 Chr.17: 4988945-4988945 on Build GRCh38
rs146330268 Chr.17: 4988943-4988943 on Build GRCh38
rs765338644 Chr.17: 4988847-4988847 on Build GRCh38
rs779648856 Chr.17: 4988947-4988947 on Build GRCh38
rs765866358 Chr.17: 4988924-4988924 on Build GRCh38
rs866131917 Chr.17: 4988865-4988865 on Build GRCh38
rs750628084 Chr.17: 4988856-4988856 on Build GRCh38
rs762262485 Chr.17: 4988838-4988838 on Build GRCh38
rs761264162 Chr.17: 4988805-4988805 on Build GRCh38
rs777228798 Chr.17: 4988959-4988959 on Build GRCh38
rs745558500 Chr.17: 4988787-4988787 on Build GRCh38
rs746534315 Chr.17: 4988948-4988948 on Build GRCh38
rs187711048 Chr.17: 4988939-4988939 on Build GRCh38
rs376708639 Chr.17: 4988887-4988887 on Build GRCh38
rs770443865 Chr.17: 4988762-4988762 on Build GRCh38
rs74744272 Chr.17: 4988881-4988881 on Build GRCh38
rs758677347 Chr.17: 4988839-4988839 on Build GRCh38
rs757043018 Chr.17: 4988906-4988906 on Build GRCh38

View Full Product Details