Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105942392-105942623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPNT
Entrez Gene ID
255743
Gene Name
nephronectin
Gene Aliases
EGFL6L, POEM
UniGene
Hs.518921
Gene Chromosome Location
Chr.4: 105895440-105971671 on Build GRCh38
Transcripts
NM_001033047.2 NM_001184690.1 NM_001184691.1 NM_001184692.1 NM_001184693.1 XM_005262888.4 XM_005262890.1 XM_011531820.2 XM_011531822.2 XM_011531823.2 XM_011531824.2 XM_011531825.2 XM_017007984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6847863 c.871G>A p.G291R Chr.4: 105942414-105942414 on Build GRCh38
1259395 c.962A>C p.K321T Chr.4: 105942505-105942505 on Build GRCh38
6728807 c.997C>T p.P333S Chr.4: 105942540-105942540 on Build GRCh38
4707845 c.866A>G p.D289G Chr.4: 105942409-105942409 on Build GRCh38
5939274 c.1042G>A p.E348K Chr.4: 105942585-105942585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143481935 Chr.4: 105942434-105942434 on Build GRCh38
rs376080328 Chr.4: 105942516-105942516 on Build GRCh38
rs767736918 Chr.4: 105942597-105942597 on Build GRCh38
rs753615257 Chr.4: 105942471-105942471 on Build GRCh38
rs755034008 Chr.4: 105942586-105942586 on Build GRCh38
rs766308266 Chr.4: 105942583-105942583 on Build GRCh38
rs756406157 Chr.4: 105942600-105942600 on Build GRCh38
rs750118953 Chr.4: 105942582-105942582 on Build GRCh38
rs765183934 Chr.4: 105942485-105942485 on Build GRCh38
rs776181375 Chr.4: 105942538-105942538 on Build GRCh38
rs182012400 Chr.4: 105942435-105942435 on Build GRCh38
rs780095462 Chr.4: 105942497-105942497 on Build GRCh38
rs769830545 Chr.4: 105942510-105942511 on Build GRCh38
rs762790628 Chr.4: 105942524-105942525 on Build GRCh38
rs768309591 Chr.4: 105942537-105942537 on Build GRCh38
rs770276744 Chr.4: 105942444-105942444 on Build GRCh38
rs748465275 Chr.4: 105942438-105942438 on Build GRCh38
rs749788166 Chr.4: 105942527-105942527 on Build GRCh38
rs760119811 Chr.4: 105942536-105942536 on Build GRCh38
rs760386796 Chr.4: 105942467-105942467 on Build GRCh38
rs149050685 Chr.4: 105942599-105942599 on Build GRCh38
rs766293318 Chr.4: 105942544-105942544 on Build GRCh38
rs764921082 Chr.4: 105942579-105942579 on Build GRCh38
rs144959760 Chr.4: 105942498-105942498 on Build GRCh38
rs899234227 Chr.4: 105942433-105942433 on Build GRCh38
rs531659716 Chr.4: 105942557-105942557 on Build GRCh38
rs369190906 Chr.4: 105942489-105942489 on Build GRCh38
rs781283871 Chr.4: 105942507-105942507 on Build GRCh38
rs146255174 Chr.4: 105942456-105942456 on Build GRCh38
rs775131445 Chr.4: 105942535-105942535 on Build GRCh38
rs540596220 Chr.4: 105942533-105942533 on Build GRCh38
rs761569898 Chr.4: 105942570-105942570 on Build GRCh38
rs780148947 Chr.4: 105942428-105942428 on Build GRCh38
rs750331445 Chr.4: 105942486-105942486 on Build GRCh38
rs115427222 Chr.4: 105942469-105942469 on Build GRCh38
rs778319042 Chr.4: 105942526-105942526 on Build GRCh38
rs747211153 Chr.4: 105942432-105942432 on Build GRCh38
rs770084338 Chr.4: 105942519-105942519 on Build GRCh38
rs773876234 Chr.4: 105942447-105942447 on Build GRCh38
rs755160066 Chr.4: 105942499-105942499 on Build GRCh38
rs368167746 Chr.4: 105942525-105942527 on Build GRCh38
rs761537863 Chr.4: 105942475-105942475 on Build GRCh38
rs139317707 Chr.4: 105942459-105942459 on Build GRCh38
rs751580473 Chr.4: 105942585-105942585 on Build GRCh38
rs750276514 Chr.4: 105942580-105942580 on Build GRCh38

View Full Product Details