Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 79942692-79942933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFTPD
Entrez Gene ID
6441
Gene Name
surfactant protein D
Gene Aliases
COLEC7, PSP-D, SFTP4, SP-D
UniGene
Hs.253495
Gene Chromosome Location
Chr.10: 79937740-79982236 on Build GRCh38
Transcripts
NM_003019.4 XM_011540087.1 XM_011540088.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139844 c.292C>T p.P98S Chr.10: 79942787-79942787 on Build GRCh38
3358881 c.250G>A p.V84I Chr.10: 79942829-79942829 on Build GRCh38
6045480 c.218G>A p.G73E Chr.10: 79942861-79942861 on Build GRCh38
3867935 c.247C>T p.P83S Chr.10: 79942832-79942832 on Build GRCh38
5593058 c.200-1G>A p.? Chr.10: 79942880-79942880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3135515 Chr.10: 79942720-79942722 on Build GRCh38
rs772740557 Chr.10: 79942728-79942728 on Build GRCh38
rs770083719 Chr.10: 79942723-79942723 on Build GRCh38
rs144218796 Chr.10: 79942891-79942891 on Build GRCh38
rs374218709 Chr.10: 79942721-79942721 on Build GRCh38
rs764018518 Chr.10: 79942765-79942765 on Build GRCh38
rs903718838 Chr.10: 79942837-79942837 on Build GRCh38
rs3135516 Chr.10: 79942746-79942746 on Build GRCh38
rs759081201 Chr.10: 79942745-79942745 on Build GRCh38
rs772924069 Chr.10: 79942883-79942883 on Build GRCh38
rs3135517 Chr.10: 79942757-79942758 on Build GRCh38
rs899830152 Chr.10: 79942911-79942911 on Build GRCh38
rs773677270 Chr.10: 79942855-79942855 on Build GRCh38
rs752623918 Chr.10: 79942750-79942750 on Build GRCh38
rs749588637 Chr.10: 79942865-79942865 on Build GRCh38
rs528953506 Chr.10: 79942902-79942902 on Build GRCh38
rs748585306 Chr.10: 79942846-79942846 on Build GRCh38
rs555493637 Chr.10: 79942717-79942717 on Build GRCh38
rs759130497 Chr.10: 79942898-79942898 on Build GRCh38
rs79085361 Chr.10: 79942841-79942841 on Build GRCh38
rs144910313 Chr.10: 79942836-79942836 on Build GRCh38
rs3135520 Chr.10: 79942821-79942821 on Build GRCh38
rs780107064 Chr.10: 79942788-79942788 on Build GRCh38
rs774357693 Chr.10: 79942913-79942913 on Build GRCh38
rs374491984 Chr.10: 79942875-79942875 on Build GRCh38
rs566166755 Chr.10: 79942786-79942786 on Build GRCh38
rs762728166 Chr.10: 79942736-79942736 on Build GRCh38
rs947977190 Chr.10: 79942824-79942824 on Build GRCh38
rs370583835 Chr.10: 79942720-79942720 on Build GRCh38
rs749220302 Chr.10: 79942729-79942729 on Build GRCh38
rs747119364 Chr.10: 79942831-79942831 on Build GRCh38
rs865817215 Chr.10: 79942851-79942851 on Build GRCh38
rs979921099 Chr.10: 79942902-79942903 on Build GRCh38
rs3135519 Chr.10: 79942759-79942760 on Build GRCh38
rs72659400 Chr.10: 79942780-79942780 on Build GRCh38
rs372491584 Chr.10: 79942743-79942743 on Build GRCh38
rs868821882 Chr.10: 79942817-79942817 on Build GRCh38
rs368425433 Chr.10: 79942737-79942737 on Build GRCh38
rs17885228 Chr.10: 79942782-79942782 on Build GRCh38
rs751087670 Chr.10: 79942779-79942780 on Build GRCh38
rs748530291 Chr.10: 79942722-79942722 on Build GRCh38

View Full Product Details