Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39062801-39063072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL5
Entrez Gene ID
51088
Gene Name
kelch like family member 5
Gene Aliases
-
UniGene
Hs.272251
Gene Chromosome Location
Chr.4: 39044796-39143101 on Build GRCh38
Transcripts
NM_001007075.2 NM_001171654.1 NM_015990.4 NM_199039.3 XM_005262655.4 XM_011513699.2 XM_011513700.2 XM_011513701.2 XM_017008273.1 XM_017008274.1 XM_017008275.1 XM_017008276.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6100023 c.442G>C p.G148R Chr.4: 39062956-39062956 on Build GRCh38
5461272 c.442G>A p.G148S Chr.4: 39062956-39062956 on Build GRCh38
6696846 c.418G>T p.E140* Chr.4: 39062932-39062932 on Build GRCh38
6360563 c.380C>A p.S127Y Chr.4: 39062894-39062894 on Build GRCh38
1055185 c.480C>T p.G160G Chr.4: 39062994-39062994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369611804 Chr.4: 39062867-39062867 on Build GRCh38
rs776566937 Chr.4: 39062910-39062910 on Build GRCh38
rs750804038 Chr.4: 39062964-39062964 on Build GRCh38
rs768959723 Chr.4: 39062887-39062887 on Build GRCh38
rs372233396 Chr.4: 39062893-39062893 on Build GRCh38
rs757645126 Chr.4: 39062956-39062956 on Build GRCh38
rs748248562 Chr.4: 39062976-39062976 on Build GRCh38
rs759531435 Chr.4: 39062912-39062912 on Build GRCh38
rs775359411 Chr.4: 39063038-39063038 on Build GRCh38
rs377278840 Chr.4: 39062995-39062995 on Build GRCh38
rs573267962 Chr.4: 39063036-39063036 on Build GRCh38
rs760104447 Chr.4: 39062916-39062916 on Build GRCh38
rs751133176 Chr.4: 39062955-39062955 on Build GRCh38
rs367628140 Chr.4: 39062831-39062831 on Build GRCh38
rs546786319 Chr.4: 39062899-39062899 on Build GRCh38
rs747451234 Chr.4: 39062833-39062833 on Build GRCh38
rs781738837 Chr.4: 39062848-39062848 on Build GRCh38
rs749372424 Chr.4: 39062882-39062882 on Build GRCh38
rs763830657 Chr.4: 39062947-39062947 on Build GRCh38
rs780673791 Chr.4: 39062969-39062969 on Build GRCh38
rs749432534 Chr.4: 39062972-39062972 on Build GRCh38
rs768567419 Chr.4: 39062974-39062974 on Build GRCh38
rs781641231 Chr.4: 39062963-39062963 on Build GRCh38
rs779223130 Chr.4: 39062975-39062975 on Build GRCh38
rs752247083 Chr.4: 39062920-39062920 on Build GRCh38
rs757516933 Chr.4: 39062841-39062841 on Build GRCh38
rs767848108 Chr.4: 39063012-39063012 on Build GRCh38
rs144755927 Chr.4: 39063027-39063027 on Build GRCh38
rs918204625 Chr.4: 39062891-39062891 on Build GRCh38
rs760537665 Chr.4: 39062895-39062895 on Build GRCh38
rs770587138 Chr.4: 39062900-39062900 on Build GRCh38
rs772329173 Chr.4: 39062987-39062987 on Build GRCh38
rs765158139 Chr.4: 39062917-39062917 on Build GRCh38
rs756751104 Chr.4: 39062968-39062968 on Build GRCh38
rs770006574 Chr.4: 39062869-39062869 on Build GRCh38
rs758139587 Chr.4: 39062829-39062829 on Build GRCh38
rs201110544 Chr.4: 39062858-39062858 on Build GRCh38
rs992306635 Chr.4: 39062879-39062879 on Build GRCh38
rs200565777 Chr.4: 39062857-39062857 on Build GRCh38
rs775769518 Chr.4: 39062871-39062871 on Build GRCh38

View Full Product Details