Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144497260-144497533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928953
Entrez Gene ID
101928953
Gene Name
uncharacterized LOC101928953
Gene Aliases
-
UniGene
Hs.521937
Gene Chromosome Location
Chr.8: 144497357-144505474 on Build GRCh38
Transcripts
XM_005272350.2 XM_011517349.2 XM_017013912.1 XM_017013913.1 XM_017013914.1 XM_017013915.1

Gene Symbol
PPP1R16A
Entrez Gene ID
84988
Gene Name
protein phosphatase 1 regulatory subunit 16A
Gene Aliases
MYPT3
UniGene
Hs.521937
Gene Chromosome Location
Chr.8: 144477987-144502121 on Build GRCh38
Transcripts
XM_005272350.2 XM_011517349.2 XM_017013912.1 XM_017013913.1 XM_017013914.1 XM_017013915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3924900 c.169G>A p.E57K Chr.8: 144497363-144497363 on Build GRCh38
4905338 c.223C>T p.L75F Chr.8: 144497417-144497417 on Build GRCh38
6434934 c.77C>A p.A26D Chr.8: 144497271-144497271 on Build GRCh38
2871771 c.91G>A p.A31T Chr.8: 144497285-144497285 on Build GRCh38
4628728 c.112G>T p.A38S Chr.8: 144497306-144497306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754912963 Chr.8: 144497410-144497412 on Build GRCh38
rs369364155 Chr.8: 144497483-144497483 on Build GRCh38
rs952186145 Chr.8: 144497373-144497373 on Build GRCh38
rs772014519 Chr.8: 144497486-144497486 on Build GRCh38
rs748876228 Chr.8: 144497479-144497479 on Build GRCh38
rs752797970 Chr.8: 144497428-144497428 on Build GRCh38
rs36115536 Chr.8: 144497488-144497489 on Build GRCh38
rs764343387 Chr.8: 144497501-144497501 on Build GRCh38
rs980366227 Chr.8: 144497484-144497484 on Build GRCh38
rs961933487 Chr.8: 144497413-144497413 on Build GRCh38
rs955227598 Chr.8: 144497359-144497359 on Build GRCh38
rs774254572 Chr.8: 144497481-144497481 on Build GRCh38
rs908474967 Chr.8: 144497379-144497379 on Build GRCh38
rs201584344 Chr.8: 144497495-144497495 on Build GRCh38
rs762602393 Chr.8: 144497278-144497278 on Build GRCh38
rs750416052 Chr.8: 144497407-144497407 on Build GRCh38
rs769115281 Chr.8: 144497469-144497469 on Build GRCh38
rs757328413 Chr.8: 144497281-144497281 on Build GRCh38
rs755220546 Chr.8: 144497298-144497298 on Build GRCh38
rs765230005 Chr.8: 144497491-144497491 on Build GRCh38
rs757572039 Chr.8: 144497511-144497511 on Build GRCh38
rs973806601 Chr.8: 144497422-144497422 on Build GRCh38
rs746384128 Chr.8: 144497460-144497460 on Build GRCh38
rs201300832 Chr.8: 144497487-144497488 on Build GRCh38
rs1029004675 Chr.8: 144497372-144497372 on Build GRCh38
rs373164819 Chr.8: 144497279-144497279 on Build GRCh38
rs867874519 Chr.8: 144497447-144497447 on Build GRCh38
rs758715057 Chr.8: 144497429-144497429 on Build GRCh38
rs745426486 Chr.8: 144497349-144497349 on Build GRCh38
rs148405688 Chr.8: 144497285-144497285 on Build GRCh38
rs376098360 Chr.8: 144497435-144497435 on Build GRCh38
rs762667060 Chr.8: 144497357-144497357 on Build GRCh38
rs780660846 Chr.8: 144497342-144497342 on Build GRCh38
rs1033386538 Chr.8: 144497345-144497345 on Build GRCh38
rs917867250 Chr.8: 144497476-144497476 on Build GRCh38
rs757735268 Chr.8: 144497440-144497440 on Build GRCh38
rs778206042 Chr.8: 144497318-144497318 on Build GRCh38
rs761818247 Chr.8: 144497384-144497384 on Build GRCh38
rs754008045 Chr.8: 144497340-144497351 on Build GRCh38
rs751972771 Chr.8: 144497436-144497436 on Build GRCh38
rs779680086 Chr.8: 144497471-144497471 on Build GRCh38
rs1019229152 Chr.8: 144497350-144497350 on Build GRCh38
rs768401691 Chr.8: 144497480-144497480 on Build GRCh38
rs769380663 Chr.8: 144497351-144497351 on Build GRCh38
rs747512091 Chr.8: 144497328-144497328 on Build GRCh38
rs752986679 Chr.8: 144497302-144497302 on Build GRCh38
rs751815377 Chr.8: 144497505-144497505 on Build GRCh38
rs142532469 Chr.8: 144497352-144497352 on Build GRCh38
rs766551858 Chr.8: 144497284-144497284 on Build GRCh38
rs767869059 Chr.8: 144497512-144497512 on Build GRCh38
rs781637549 Chr.8: 144497453-144497453 on Build GRCh38
rs201796418 Chr.8: 144497316-144497316 on Build GRCh38
rs771488119 Chr.8: 144497333-144497333 on Build GRCh38
rs759486140 Chr.8: 144497408-144497408 on Build GRCh38
rs936390563 Chr.8: 144497516-144497516 on Build GRCh38
rs778961846 Chr.8: 144497300-144497300 on Build GRCh38
rs1029522539 Chr.8: 144497358-144497358 on Build GRCh38
rs145969525 Chr.8: 144497375-144497375 on Build GRCh38
rs774344463 Chr.8: 144497383-144497383 on Build GRCh38
rs767603187 Chr.8: 144497404-144497404 on Build GRCh38
rs200756116 Chr.8: 144497280-144497280 on Build GRCh38

View Full Product Details