Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90085278-90085503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514509 c.977C>T p.S326F Chr.15: 90085378-90085378 on Build GRCh38
6494434 c.1050C>T p.T350T Chr.15: 90085305-90085305 on Build GRCh38
6318877 c.1050C>A p.T350T Chr.15: 90085305-90085305 on Build GRCh38
1658762 c.1054A>C p.T352P Chr.15: 90085301-90085301 on Build GRCh38
6954337 c.1077G>C p.Q359H Chr.15: 90085278-90085278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61737002 Chr.15: 90085362-90085362 on Build GRCh38
rs61737003 Chr.15: 90085359-90085359 on Build GRCh38
rs567288602 Chr.15: 90085420-90085420 on Build GRCh38
rs758449799 Chr.15: 90085395-90085395 on Build GRCh38
rs897159348 Chr.15: 90085336-90085336 on Build GRCh38
rs777753317 Chr.15: 90085402-90085402 on Build GRCh38
rs11540478 Chr.15: 90085305-90085305 on Build GRCh38
rs766516876 Chr.15: 90085365-90085365 on Build GRCh38
rs769180013 Chr.15: 90085335-90085335 on Build GRCh38
rs190078206 Chr.15: 90085317-90085317 on Build GRCh38
rs531222215 Chr.15: 90085426-90085426 on Build GRCh38
rs140596855 Chr.15: 90085352-90085352 on Build GRCh38
rs770548230 Chr.15: 90085301-90085301 on Build GRCh38
rs974929266 Chr.15: 90085469-90085469 on Build GRCh38
rs751178080 Chr.15: 90085395-90085396 on Build GRCh38
rs1018434130 Chr.15: 90085399-90085399 on Build GRCh38
rs745476180 Chr.15: 90085433-90085433 on Build GRCh38
rs765909746 Chr.15: 90085303-90085304 on Build GRCh38
rs774623780 Chr.15: 90085343-90085343 on Build GRCh38
rs776344212 Chr.15: 90085303-90085303 on Build GRCh38
rs143339918 Chr.15: 90085296-90085296 on Build GRCh38
rs757012102 Chr.15: 90085421-90085421 on Build GRCh38
rs927842567 Chr.15: 90085330-90085330 on Build GRCh38
rs564700723 Chr.15: 90085345-90085345 on Build GRCh38
rs767061831 Chr.15: 90085316-90085316 on Build GRCh38
rs943106383 Chr.15: 90085412-90085412 on Build GRCh38
rs370350675 Chr.15: 90085332-90085332 on Build GRCh38
rs752711856 Chr.15: 90085368-90085368 on Build GRCh38
rs550377664 Chr.15: 90085370-90085370 on Build GRCh38
rs561368494 Chr.15: 90085444-90085444 on Build GRCh38
rs61733005 Chr.15: 90085358-90085358 on Build GRCh38
rs745389409 Chr.15: 90085304-90085304 on Build GRCh38
rs375907103 Chr.15: 90085430-90085430 on Build GRCh38
rs1036260002 Chr.15: 90085333-90085333 on Build GRCh38

View Full Product Details