Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64927897-64928150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R5B
Entrez Gene ID
5526
Gene Name
protein phosphatase 2 regulatory subunit B'beta
Gene Aliases
B56B, PR61B
UniGene
Hs.75199
Gene Chromosome Location
Chr.11: 64917553-64934478 on Build GRCh38
Transcripts
NM_006244.3 XM_011545132.2 XM_011545133.1 XM_011545134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2164795 c.553G>A p.V185M Chr.11: 64928120-64928120 on Build GRCh38
7125031 c.561G>C p.K187N Chr.11: 64928128-64928128 on Build GRCh38
3739216 c.552C>T p.S184S Chr.11: 64928119-64928119 on Build GRCh38
930265 c.574C>A p.Q192K Chr.11: 64928141-64928141 on Build GRCh38
930264 c.565T>C p.Y189H Chr.11: 64928132-64928132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371043782 Chr.11: 64927951-64927951 on Build GRCh38
rs1001632475 Chr.11: 64928061-64928061 on Build GRCh38
rs374171324 Chr.11: 64928040-64928040 on Build GRCh38
rs267603109 Chr.11: 64928083-64928083 on Build GRCh38
rs780240927 Chr.11: 64928040-64928041 on Build GRCh38
rs757729823 Chr.11: 64927999-64927999 on Build GRCh38
rs898688637 Chr.11: 64928097-64928097 on Build GRCh38
rs372825084 Chr.11: 64927924-64927924 on Build GRCh38
rs757705043 Chr.11: 64928098-64928098 on Build GRCh38
rs763065116 Chr.11: 64927922-64927922 on Build GRCh38
rs1053044386 Chr.11: 64927983-64927983 on Build GRCh38
rs150933072 Chr.11: 64928088-64928088 on Build GRCh38
rs746347844 Chr.11: 64928116-64928116 on Build GRCh38
rs995688391 Chr.11: 64928101-64928101 on Build GRCh38
rs1024138989 Chr.11: 64928030-64928030 on Build GRCh38
rs535137383 Chr.11: 64928018-64928018 on Build GRCh38
rs775390875 Chr.11: 64928120-64928120 on Build GRCh38
rs376545618 Chr.11: 64927943-64927943 on Build GRCh38
rs752289539 Chr.11: 64928049-64928049 on Build GRCh38
rs781732494 Chr.11: 64928100-64928100 on Build GRCh38
rs147663616 Chr.11: 64928087-64928087 on Build GRCh38
rs749528896 Chr.11: 64928047-64928047 on Build GRCh38
rs753767825 Chr.11: 64928025-64928025 on Build GRCh38
rs923884833 Chr.11: 64927957-64927957 on Build GRCh38
rs754996569 Chr.11: 64928036-64928036 on Build GRCh38
rs935329052 Chr.11: 64927973-64927973 on Build GRCh38
rs757950819 Chr.11: 64928062-64928062 on Build GRCh38
rs770383136 Chr.11: 64928119-64928119 on Build GRCh38

View Full Product Details