Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140809278-140809551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_031500.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3239953 c.2158G>A p.A720T Chr.5: 140809345-140809345 on Build GRCh38
3918813 c.2353C>T p.Q785* Chr.5: 140809540-140809540 on Build GRCh38
3239966 c.2253G>A p.S751S Chr.5: 140809440-140809440 on Build GRCh38
332053 c.2159C>T p.A720V Chr.5: 140809346-140809346 on Build GRCh38
1130909 c.2157C>T p.T719T Chr.5: 140809344-140809344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781909645 Chr.5: 140809400-140809400 on Build GRCh38
rs782200079 Chr.5: 140809410-140809410 on Build GRCh38
rs782500159 Chr.5: 140809495-140809495 on Build GRCh38
rs782810628 Chr.5: 140809491-140809491 on Build GRCh38
rs782392679 Chr.5: 140809366-140809366 on Build GRCh38
rs138998178 Chr.5: 140809345-140809345 on Build GRCh38
rs541059549 Chr.5: 140809414-140809414 on Build GRCh38
rs115463013 Chr.5: 140809401-140809401 on Build GRCh38
rs782399688 Chr.5: 140809366-140809366 on Build GRCh38
rs782243123 Chr.5: 140809431-140809431 on Build GRCh38
rs782466044 Chr.5: 140809339-140809339 on Build GRCh38
rs781955144 Chr.5: 140809440-140809440 on Build GRCh38
rs760109122 Chr.5: 140809433-140809433 on Build GRCh38
rs543555386 Chr.5: 140809367-140809367 on Build GRCh38
rs782154101 Chr.5: 140809372-140809372 on Build GRCh38
rs782650472 Chr.5: 140809503-140809503 on Build GRCh38
rs148640226 Chr.5: 140809438-140809438 on Build GRCh38
rs781828358 Chr.5: 140809405-140809405 on Build GRCh38
rs782561720 Chr.5: 140809409-140809409 on Build GRCh38
rs558389511 Chr.5: 140809383-140809383 on Build GRCh38
rs576165362 Chr.5: 140809337-140809337 on Build GRCh38
rs782146620 Chr.5: 140809446-140809446 on Build GRCh38
rs782745865 Chr.5: 140809475-140809475 on Build GRCh38
rs782554104 Chr.5: 140809508-140809508 on Build GRCh38
rs782610697 Chr.5: 140809344-140809344 on Build GRCh38
rs782158465 Chr.5: 140809319-140809319 on Build GRCh38
rs782036778 Chr.5: 140809334-140809334 on Build GRCh38
rs781959582 Chr.5: 140809476-140809476 on Build GRCh38
rs374214403 Chr.5: 140809511-140809511 on Build GRCh38
rs782718692 Chr.5: 140809305-140809305 on Build GRCh38
rs781869245 Chr.5: 140809333-140809333 on Build GRCh38
rs145922941 Chr.5: 140809415-140809415 on Build GRCh38
rs781791208 Chr.5: 140809476-140809476 on Build GRCh38
rs782212508 Chr.5: 140809505-140809505 on Build GRCh38
rs782047510 Chr.5: 140809392-140809392 on Build GRCh38
rs17844285 Chr.5: 140809306-140809306 on Build GRCh38
rs781936039 Chr.5: 140809371-140809371 on Build GRCh38
rs781832159 Chr.5: 140809389-140809389 on Build GRCh38
rs782448560 Chr.5: 140809477-140809477 on Build GRCh38
rs782451445 Chr.5: 140809350-140809350 on Build GRCh38
rs145154617 Chr.5: 140809323-140809323 on Build GRCh38
rs782714901 Chr.5: 140809388-140809388 on Build GRCh38
rs373952073 Chr.5: 140809404-140809404 on Build GRCh38
rs782496896 Chr.5: 140809406-140809406 on Build GRCh38
rs781993408 Chr.5: 140809528-140809528 on Build GRCh38
rs150783892 Chr.5: 140809474-140809474 on Build GRCh38
rs782248893 Chr.5: 140809362-140809362 on Build GRCh38
rs782270940 Chr.5: 140809529-140809529 on Build GRCh38
rs782009545 Chr.5: 140809462-140809462 on Build GRCh38
rs782686583 Chr.5: 140809361-140809361 on Build GRCh38
rs782301046 Chr.5: 140809445-140809445 on Build GRCh38
rs77547730 Chr.5: 140809378-140809378 on Build GRCh38
rs146943849 Chr.5: 140809455-140809455 on Build GRCh38
rs370062696 Chr.5: 140809377-140809377 on Build GRCh38
rs781910272 Chr.5: 140809494-140809494 on Build GRCh38

View Full Product Details