Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45921991-45922252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAF11
Entrez Gene ID
9169
Gene Name
SR-related CTD associated factor 11
Gene Aliases
CASP11, SFRS2IP, SIP1, SRRP129, SRSF2IP
UniGene
Hs.210367
Gene Chromosome Location
Chr.12: 45919131-45992040 on Build GRCh38
Transcripts
NM_004719.2 XM_005269230.2 XM_011538984.2 XM_011538985.1 XM_017020217.1 XM_017020218.1 XM_017020219.1 XM_017020220.1 XM_017020221.1 XM_017020222.1 XM_017020223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6772341 c.4282A>G p.K1428E Chr.12: 45922158-45922158 on Build GRCh38
4547617 c.4270G>T p.V1424L Chr.12: 45922170-45922170 on Build GRCh38
6772329 c.4308T>C p.Y1436Y Chr.12: 45922132-45922132 on Build GRCh38
6872424 c.4335G>A p.K1445K Chr.12: 45922105-45922105 on Build GRCh38
6772327 c.4351delA p.T1451fs*12 Chr.12: 45922089-45922089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971577357 Chr.12: 45922206-45922206 on Build GRCh38
rs375184453 Chr.12: 45922028-45922028 on Build GRCh38
rs754816885 Chr.12: 45922207-45922207 on Build GRCh38
rs771679383 Chr.12: 45922037-45922037 on Build GRCh38
rs747503732 Chr.12: 45922042-45922042 on Build GRCh38
rs752376610 Chr.12: 45922213-45922213 on Build GRCh38
rs766646359 Chr.12: 45922130-45922130 on Build GRCh38
rs771235077 Chr.12: 45922056-45922056 on Build GRCh38
rs376431197 Chr.12: 45922135-45922135 on Build GRCh38
rs753724763 Chr.12: 45922204-45922204 on Build GRCh38
rs765024624 Chr.12: 45922210-45922210 on Build GRCh38
rs762806362 Chr.12: 45922205-45922206 on Build GRCh38
rs753436067 Chr.12: 45922026-45922026 on Build GRCh38
rs766100394 Chr.12: 45922184-45922184 on Build GRCh38
rs911060635 Chr.12: 45922216-45922216 on Build GRCh38
rs372902559 Chr.12: 45922086-45922086 on Build GRCh38
rs778695189 Chr.12: 45922031-45922031 on Build GRCh38
rs780612038 Chr.12: 45922148-45922148 on Build GRCh38
rs760594689 Chr.12: 45922161-45922161 on Build GRCh38
rs973884387 Chr.12: 45922051-45922051 on Build GRCh38
rs985222935 Chr.12: 45922187-45922187 on Build GRCh38
rs879073731 Chr.12: 45922214-45922214 on Build GRCh38
rs747729493 Chr.12: 45922034-45922034 on Build GRCh38
rs749966940 Chr.12: 45922167-45922167 on Build GRCh38
rs779858245 Chr.12: 45922018-45922018 on Build GRCh38
rs200034095 Chr.12: 45922038-45922038 on Build GRCh38
rs770065046 Chr.12: 45922079-45922079 on Build GRCh38
rs775655813 Chr.12: 45922109-45922109 on Build GRCh38
rs776991680 Chr.12: 45922068-45922068 on Build GRCh38
rs920664231 Chr.12: 45922103-45922103 on Build GRCh38
rs139826670 Chr.12: 45922159-45922159 on Build GRCh38
rs917016159 Chr.12: 45922143-45922143 on Build GRCh38
rs746200797 Chr.12: 45922077-45922077 on Build GRCh38
rs758007686 Chr.12: 45922220-45922220 on Build GRCh38

View Full Product Details