Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82820908-82821181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC31A
Entrez Gene ID
22872
Gene Name
SEC31 homolog A, COPII coat complex component
Gene Aliases
ABP125, ABP130, HSPC275, HSPC334, SEC31L1
UniGene
Hs.370024
Gene Chromosome Location
Chr.4: 82818509-82900571 on Build GRCh38
Transcripts
NM_001077206.3 NM_001077207.3 NM_001077208.3 NM_001191049.2 NM_001300744.2 NM_001300745.2 NM_001318119.1 NM_001318120.1 NM_016211.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584045 c.3473G>A p.R1158K Chr.4: 82821047-82821047 on Build GRCh38
1328375 c.3459G>C p.L1153L Chr.4: 82821061-82821061 on Build GRCh38
5459121 c.3412-6C>A p.? Chr.4: 82821114-82821114 on Build GRCh38
5584046 c.3512G>A p.R1171K Chr.4: 82821047-82821047 on Build GRCh38
5768659 c.3496C>T p.L1166L Chr.4: 82821063-82821063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777787340 Chr.4: 82821144-82821144 on Build GRCh38
rs761429193 Chr.4: 82821097-82821097 on Build GRCh38
rs753159650 Chr.4: 82821129-82821129 on Build GRCh38
rs375844952 Chr.4: 82820983-82820983 on Build GRCh38
rs559358269 Chr.4: 82820969-82820969 on Build GRCh38
rs370315907 Chr.4: 82821104-82821104 on Build GRCh38
rs917019603 Chr.4: 82820944-82820944 on Build GRCh38
rs921902117 Chr.4: 82821014-82821014 on Build GRCh38
rs550598763 Chr.4: 82821055-82821055 on Build GRCh38
rs748219050 Chr.4: 82821004-82821004 on Build GRCh38
rs763515528 Chr.4: 82821072-82821072 on Build GRCh38
rs192746270 Chr.4: 82821010-82821010 on Build GRCh38
rs775307375 Chr.4: 82821051-82821051 on Build GRCh38
rs768695514 Chr.4: 82821150-82821150 on Build GRCh38
rs549237449 Chr.4: 82820935-82820935 on Build GRCh38
rs769677265 Chr.4: 82821037-82821037 on Build GRCh38
rs983130011 Chr.4: 82820949-82820949 on Build GRCh38
rs1048998568 Chr.4: 82821081-82821081 on Build GRCh38
rs930451853 Chr.4: 82821075-82821075 on Build GRCh38
rs774048176 Chr.4: 82821074-82821074 on Build GRCh38
rs773953463 Chr.4: 82820998-82820999 on Build GRCh38
rs746135958 Chr.4: 82821083-82821083 on Build GRCh38
rs766826750 Chr.4: 82821103-82821103 on Build GRCh38
rs745771275 Chr.4: 82821035-82821035 on Build GRCh38
rs571414144 Chr.4: 82821135-82821135 on Build GRCh38
rs184276133 Chr.4: 82821061-82821061 on Build GRCh38
rs770711117 Chr.4: 82821018-82821018 on Build GRCh38
rs376326449 Chr.4: 82821020-82821020 on Build GRCh38
rs755425156 Chr.4: 82821116-82821116 on Build GRCh38
rs373388778 Chr.4: 82821126-82821126 on Build GRCh38
rs758431672 Chr.4: 82821007-82821007 on Build GRCh38

View Full Product Details