Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175947251-175947520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LNPK
Entrez Gene ID
80856
Gene Name
lunapark, ER junction formation factor
Gene Aliases
KIAA1715, LNP, LNP1, Ul, ulnaless
UniGene
Hs.209561
Gene Chromosome Location
Chr.2: 175923862-176002901 on Build GRCh38
Transcripts
NM_001305008.1 NM_001305009.1 NM_001305010.1 NM_001305011.1 NM_030650.2 NR_130941.1 XM_005246878.3 XM_005246879.4 XM_005246880.3 XM_006712783.2 XM_011511945.2 XM_017005055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009819 c.673C>A p.L225I Chr.2: 175947513-175947513 on Build GRCh38
4866024 c.673C>A p.L225I Chr.2: 175947513-175947513 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948313312 Chr.2: 175947391-175947391 on Build GRCh38
rs749580123 Chr.2: 175947448-175947448 on Build GRCh38
rs751713236 Chr.2: 175947418-175947418 on Build GRCh38
rs1052502944 Chr.2: 175947444-175947444 on Build GRCh38
rs902041490 Chr.2: 175947370-175947370 on Build GRCh38
rs1035505038 Chr.2: 175947473-175947473 on Build GRCh38
rs139257549 Chr.2: 175947419-175947419 on Build GRCh38
rs925514474 Chr.2: 175947306-175947306 on Build GRCh38
rs920809418 Chr.2: 175947362-175947362 on Build GRCh38
rs757807998 Chr.2: 175947482-175947482 on Build GRCh38
rs939435775 Chr.2: 175947428-175947428 on Build GRCh38
rs940868008 Chr.2: 175947327-175947327 on Build GRCh38
rs764813967 Chr.2: 175947451-175947451 on Build GRCh38
rs753558236 Chr.2: 175947436-175947436 on Build GRCh38
rs933397046 Chr.2: 175947372-175947372 on Build GRCh38
rs185144165 Chr.2: 175947426-175947426 on Build GRCh38
rs143694399 Chr.2: 175947460-175947460 on Build GRCh38
rs577691489 Chr.2: 175947336-175947336 on Build GRCh38
rs544085332 Chr.2: 175947492-175947492 on Build GRCh38
rs973580068 Chr.2: 175947335-175947335 on Build GRCh38
rs905751745 Chr.2: 175947458-175947458 on Build GRCh38
rs886528559 Chr.2: 175947405-175947405 on Build GRCh38
rs908023793 Chr.2: 175947284-175947284 on Build GRCh38
rs1022085350 Chr.2: 175947483-175947483 on Build GRCh38
rs553028692 Chr.2: 175947300-175947300 on Build GRCh38

View Full Product Details