Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238082026-238082285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCLY
Entrez Gene ID
51540
Gene Name
selenocysteine lyase
Gene Aliases
SCL, hSCL
UniGene
Hs.731909
Gene Chromosome Location
Chr.2: 238060924-238099413 on Build GRCh38
Transcripts
NM_016510.5 NR_037904.1

Gene Symbol
UBE2F-SCLY
Entrez Gene ID
100533179
Gene Name
UBE2F-SCLY readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.471785
Gene Chromosome Location
Chr.2: 237966945-238099413 on Build GRCh38
Transcripts
NM_016510.5 NR_037904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364462 c.730C>T p.Q244* Chr.2: 238082138-238082138 on Build GRCh38
5396586 c.681C>T p.P227P Chr.2: 238082113-238082113 on Build GRCh38
145908 c.667G>A p.A223T Chr.2: 238082075-238082075 on Build GRCh38
1406686 c.682C>T p.R228W Chr.2: 238082090-238082090 on Build GRCh38
1406687 c.748C>T p.R250C Chr.2: 238082156-238082156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367556253 Chr.2: 238082120-238082120 on Build GRCh38
rs199950944 Chr.2: 238082251-238082251 on Build GRCh38
rs929448495 Chr.2: 238082171-238082171 on Build GRCh38
rs769034504 Chr.2: 238082059-238082059 on Build GRCh38
rs759790678 Chr.2: 238082091-238082091 on Build GRCh38
rs772254976 Chr.2: 238082138-238082138 on Build GRCh38
rs773584417 Chr.2: 238082067-238082067 on Build GRCh38
rs751386732 Chr.2: 238082116-238082116 on Build GRCh38
rs745554562 Chr.2: 238082121-238082121 on Build GRCh38
rs145727476 Chr.2: 238082165-238082165 on Build GRCh38
rs760808689 Chr.2: 238082069-238082069 on Build GRCh38
rs1002290489 Chr.2: 238082250-238082250 on Build GRCh38
rs762674439 Chr.2: 238082128-238082128 on Build GRCh38
rs749987780 Chr.2: 238082197-238082197 on Build GRCh38
rs150339508 Chr.2: 238082119-238082119 on Build GRCh38
rs754109026 Chr.2: 238082090-238082090 on Build GRCh38
rs755611829 Chr.2: 238082198-238082198 on Build GRCh38
rs764157314 Chr.2: 238082206-238082206 on Build GRCh38
rs939001178 Chr.2: 238082137-238082137 on Build GRCh38
rs748877878 Chr.2: 238082254-238082254 on Build GRCh38
rs748573336 Chr.2: 238082133-238082133 on Build GRCh38
rs761036731 Chr.2: 238082153-238082153 on Build GRCh38
rs190876087 Chr.2: 238082255-238082255 on Build GRCh38
rs367874650 Chr.2: 238082239-238082239 on Build GRCh38
rs773779138 Chr.2: 238082147-238082147 on Build GRCh38
rs768478627 Chr.2: 238082159-238082159 on Build GRCh38
rs765523812 Chr.2: 238082093-238082093 on Build GRCh38
rs868676086 Chr.2: 238082140-238082140 on Build GRCh38
rs147560427 Chr.2: 238082046-238082046 on Build GRCh38
rs151214628 Chr.2: 238082075-238082075 on Build GRCh38
rs752963333 Chr.2: 238082156-238082156 on Build GRCh38
rs750251393 Chr.2: 238082155-238082155 on Build GRCh38
rs1010944021 Chr.2: 238082179-238082179 on Build GRCh38
rs758253213 Chr.2: 238082098-238082098 on Build GRCh38
rs372090115 Chr.2: 238082127-238082127 on Build GRCh38
rs992409129 Chr.2: 238082139-238082139 on Build GRCh38
rs762846716 Chr.2: 238082157-238082157 on Build GRCh38
rs944634348 Chr.2: 238082158-238082158 on Build GRCh38
rs752588917 Chr.2: 238082097-238082097 on Build GRCh38
rs774661539 Chr.2: 238082060-238082060 on Build GRCh38
rs375406062 Chr.2: 238082175-238082175 on Build GRCh38
rs759664845 Chr.2: 238082154-238082154 on Build GRCh38
rs142210384 Chr.2: 238082066-238082066 on Build GRCh38
rs779681464 Chr.2: 238082225-238082225 on Build GRCh38
rs777834921 Chr.2: 238082115-238082115 on Build GRCh38
rs138193153 Chr.2: 238082180-238082180 on Build GRCh38
rs879042936 Chr.2: 238082240-238082240 on Build GRCh38

View Full Product Details