Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49100415-49100674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBR1L
Entrez Gene ID
55716
Gene Name
limb development membrane protein 1 like
Gene Aliases
LIMR
UniGene
Hs.272838
Gene Chromosome Location
Chr.12: 49096551-49111535 on Build GRCh38
Transcripts
NM_001300750.1 NM_001300751.1 NM_018113.3 XM_011538563.2 XM_011538564.2 XM_011538565.2 XM_011538566.2 XM_017019626.1 XM_017019627.1 XM_017019628.1 XM_017019629.1 XM_017019630.1 XM_017019631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6701704 c.1100A>G p.Y367C Chr.12: 49100614-49100614 on Build GRCh38
4042424 c.1194C>T p.S398S Chr.12: 49100419-49100419 on Build GRCh38
84158 c.1068-4G>A p.? Chr.12: 49100650-49100650 on Build GRCh38
1362072 c.1141C>T p.R381W Chr.12: 49100588-49100588 on Build GRCh38
1362073 c.1126C>T p.R376W Chr.12: 49100588-49100588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769611662 Chr.12: 49100521-49100521 on Build GRCh38
rs936843426 Chr.12: 49100468-49100468 on Build GRCh38
rs752375877 Chr.12: 49100506-49100506 on Build GRCh38
rs201126566 Chr.12: 49100597-49100597 on Build GRCh38
rs138195319 Chr.12: 49100521-49100521 on Build GRCh38
rs377091515 Chr.12: 49100560-49100560 on Build GRCh38
rs781225062 Chr.12: 49100438-49100439 on Build GRCh38
rs369964677 Chr.12: 49100445-49100445 on Build GRCh38
rs762089902 Chr.12: 49100467-49100467 on Build GRCh38
rs374969605 Chr.12: 49100475-49100475 on Build GRCh38
rs745769683 Chr.12: 49100526-49100526 on Build GRCh38
rs533310920 Chr.12: 49100497-49100497 on Build GRCh38
rs757994793 Chr.12: 49100519-49100519 on Build GRCh38
rs770839198 Chr.12: 49100454-49100454 on Build GRCh38
rs753567440 Chr.12: 49100457-49100457 on Build GRCh38
rs774526219 Chr.12: 49100543-49100543 on Build GRCh38
rs761091914 Chr.12: 49100484-49100484 on Build GRCh38
rs768895149 Chr.12: 49100527-49100527 on Build GRCh38
rs764745912 Chr.12: 49100588-49100588 on Build GRCh38
rs758046116 Chr.12: 49100596-49100596 on Build GRCh38
rs775437707 Chr.12: 49100461-49100461 on Build GRCh38
rs748079152 Chr.12: 49100457-49100464 on Build GRCh38
rs1052735366 Chr.12: 49100650-49100650 on Build GRCh38
rs745841941 Chr.12: 49100639-49100639 on Build GRCh38
rs377594070 Chr.12: 49100573-49100573 on Build GRCh38
rs892732463 Chr.12: 49100487-49100487 on Build GRCh38
rs777237286 Chr.12: 49100576-49100576 on Build GRCh38
rs200270392 Chr.12: 49100545-49100545 on Build GRCh38
rs751157694 Chr.12: 49100616-49100616 on Build GRCh38
rs948203808 Chr.12: 49100524-49100524 on Build GRCh38
rs369461902 Chr.12: 49100586-49100586 on Build GRCh38
rs563804033 Chr.12: 49100522-49100522 on Build GRCh38
rs766980443 Chr.12: 49100494-49100494 on Build GRCh38
rs761163099 Chr.12: 49100564-49100564 on Build GRCh38
rs756059271 Chr.12: 49100640-49100640 on Build GRCh38
rs371663533 Chr.12: 49100471-49100471 on Build GRCh38
rs772354061 Chr.12: 49100559-49100559 on Build GRCh38
rs777506496 Chr.12: 49100455-49100455 on Build GRCh38
rs534094031 Chr.12: 49100634-49100634 on Build GRCh38
rs773051462 Chr.12: 49100545-49100546 on Build GRCh38
rs752140938 Chr.12: 49100589-49100589 on Build GRCh38
rs995312446 Chr.12: 49100587-49100587 on Build GRCh38
rs763159593 Chr.12: 49100464-49100464 on Build GRCh38
rs781247529 Chr.12: 49100523-49100523 on Build GRCh38
rs774761434 Chr.12: 49100466-49100466 on Build GRCh38
rs891479202 Chr.12: 49100652-49100652 on Build GRCh38
rs373615407 Chr.12: 49100569-49100569 on Build GRCh38
rs768604312 Chr.12: 49100465-49100465 on Build GRCh38
rs757123145 Chr.12: 49100618-49100618 on Build GRCh38
rs778366312 Chr.12: 49100503-49100503 on Build GRCh38
rs780817098 Chr.12: 49100635-49100635 on Build GRCh38
rs370212450 Chr.12: 49100579-49100579 on Build GRCh38
rs117771708 Chr.12: 49100549-49100549 on Build GRCh38
rs763803260 Chr.12: 49100614-49100614 on Build GRCh38

View Full Product Details