Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24181334-24181581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
UniGene
Hs.411865
Gene Chromosome Location
Chr.14: 24180219-24188918 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
327450 c.2994G>A p.W998* Chr.14: 24181564-24181564 on Build GRCh38
3983205 c.2980G>A p.D994N Chr.14: 24181578-24181578 on Build GRCh38
1369313 c.3008G>A p.R1003H Chr.14: 24181550-24181550 on Build GRCh38
2032753 c.3002T>C p.I1001T Chr.14: 24181556-24181556 on Build GRCh38
7303596 c.3042C>A p.D1014E Chr.14: 24181516-24181516 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1025464324 Chr.14: 24181490-24181490 on Build GRCh38
rs1000580318 Chr.14: 24181495-24181495 on Build GRCh38
rs772527999 Chr.14: 24181472-24181474 on Build GRCh38
rs890284402 Chr.14: 24181435-24181435 on Build GRCh38
rs549537225 Chr.14: 24181479-24181479 on Build GRCh38
rs372609334 Chr.14: 24181516-24181516 on Build GRCh38
rs958410864 Chr.14: 24181551-24181551 on Build GRCh38
rs1017459968 Chr.14: 24181457-24181457 on Build GRCh38
rs753015395 Chr.14: 24181555-24181555 on Build GRCh38
rs143309918 Chr.14: 24181420-24181420 on Build GRCh38
rs772678362 Chr.14: 24181505-24181505 on Build GRCh38
rs756375329 Chr.14: 24181556-24181556 on Build GRCh38
rs898882460 Chr.14: 24181375-24181375 on Build GRCh38
rs199631729 Chr.14: 24181543-24181543 on Build GRCh38
rs762679264 Chr.14: 24181478-24181495 on Build GRCh38
rs758816055 Chr.14: 24181386-24181386 on Build GRCh38
rs148297441 Chr.14: 24181456-24181456 on Build GRCh38
rs1037696977 Chr.14: 24181441-24181441 on Build GRCh38
rs746236235 Chr.14: 24181377-24181377 on Build GRCh38
rs993592304 Chr.14: 24181489-24181489 on Build GRCh38
rs778034805 Chr.14: 24181558-24181558 on Build GRCh38
rs1037473681 Chr.14: 24181372-24181372 on Build GRCh38
rs369101802 Chr.14: 24181550-24181550 on Build GRCh38
rs964762436 Chr.14: 24181474-24181474 on Build GRCh38
rs143030433 Chr.14: 24181548-24181548 on Build GRCh38
rs889141012 Chr.14: 24181410-24181410 on Build GRCh38
rs933568494 Chr.14: 24181389-24181389 on Build GRCh38
rs539625985 Chr.14: 24181478-24181478 on Build GRCh38
rs369576079 Chr.14: 24181509-24181509 on Build GRCh38
rs549450603 Chr.14: 24181393-24181393 on Build GRCh38

View Full Product Details