Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71986929-71987186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF121
Entrez Gene ID
55298
Gene Name
ring finger protein 121
Gene Aliases
-
UniGene
Hs.368554
Gene Chromosome Location
Chr.11: 71928701-71997597 on Build GRCh38
Transcripts
NM_001300926.1 NM_018320.4 NR_024147.1 NR_024148.1 XM_006718629.1 XM_006718630.1 XM_006718632.1 XM_011545150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6014555 c.399-5C>T p.? Chr.11: 71986999-71986999 on Build GRCh38
7426931 c.414G>C p.W138C Chr.11: 71987019-71987019 on Build GRCh38
7110234 c.426C>T p.I142I Chr.11: 71987031-71987031 on Build GRCh38
7448881 c.469G>T p.V157F Chr.11: 71987074-71987074 on Build GRCh38
6767244 c.490C>A p.L164I Chr.11: 71987095-71987095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751991383 Chr.11: 71986968-71986969 on Build GRCh38
rs375681453 Chr.11: 71986959-71986959 on Build GRCh38
rs1050717870 Chr.11: 71986962-71986962 on Build GRCh38
rs752840273 Chr.11: 71987085-71987085 on Build GRCh38
rs755318917 Chr.11: 71987077-71987077 on Build GRCh38
rs773980863 Chr.11: 71987032-71987032 on Build GRCh38
rs892105046 Chr.11: 71986964-71986965 on Build GRCh38
rs1011793762 Chr.11: 71987005-71987005 on Build GRCh38
rs533299668 Chr.11: 71986974-71986974 on Build GRCh38
rs945130478 Chr.11: 71986972-71986972 on Build GRCh38
rs747510704 Chr.11: 71987104-71987104 on Build GRCh38
rs757616109 Chr.11: 71987105-71987105 on Build GRCh38
rs762807606 Chr.11: 71987023-71987023 on Build GRCh38
rs767269061 Chr.11: 71987054-71987054 on Build GRCh38
rs868704947 Chr.11: 71987072-71987072 on Build GRCh38
rs758925411 Chr.11: 71987089-71987089 on Build GRCh38
rs769277637 Chr.11: 71986993-71986993 on Build GRCh38
rs770450092 Chr.11: 71986965-71986965 on Build GRCh38
rs1022904572 Chr.11: 71987021-71987021 on Build GRCh38
rs745551302 Chr.11: 71986992-71986992 on Build GRCh38
rs765341855 Chr.11: 71987079-71987079 on Build GRCh38
rs775044183 Chr.11: 71987006-71987006 on Build GRCh38
rs761641468 Chr.11: 71987045-71987045 on Build GRCh38
rs764042875 Chr.11: 71987031-71987031 on Build GRCh38
rs551382491 Chr.11: 71987068-71987068 on Build GRCh38
rs373014603 Chr.11: 71987157-71987157 on Build GRCh38
rs757179442 Chr.11: 71986957-71986957 on Build GRCh38
rs774935466 Chr.11: 71987136-71987136 on Build GRCh38
rs778454103 Chr.11: 71987090-71987090 on Build GRCh38
rs748830769 Chr.11: 71987148-71987148 on Build GRCh38
rs981343293 Chr.11: 71987080-71987080 on Build GRCh38
rs146102089 Chr.11: 71987109-71987109 on Build GRCh38
rs369775219 Chr.11: 71987124-71987124 on Build GRCh38
rs569635385 Chr.11: 71987111-71987111 on Build GRCh38
rs536601275 Chr.11: 71987121-71987121 on Build GRCh38

View Full Product Details