Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62593876-62594131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTA2
Entrez Gene ID
9219
Gene Name
metastasis associated 1 family member 2
Gene Aliases
MTA1L1, PID
UniGene
Hs.173043
Gene Chromosome Location
Chr.11: 62593203-62601840 on Build GRCh38
Transcripts
NM_004739.3 XM_017018561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716131 c.1849C>T p.R617W Chr.11: 62594033-62594033 on Build GRCh38
240733 c.1843G>C p.A615P Chr.11: 62594039-62594039 on Build GRCh38
6000037 c.1889G>A p.R630H Chr.11: 62593993-62593993 on Build GRCh38
929715 c.1994T>A p.V665D Chr.11: 62593888-62593888 on Build GRCh38
4034789 c.1853A>G p.K618R Chr.11: 62594029-62594029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373340730 Chr.11: 62593978-62593978 on Build GRCh38
rs772302275 Chr.11: 62593977-62593977 on Build GRCh38
rs766071392 Chr.11: 62593938-62593939 on Build GRCh38
rs113716257 Chr.11: 62593910-62593910 on Build GRCh38
rs578054510 Chr.11: 62593913-62593913 on Build GRCh38
rs1027219892 Chr.11: 62594105-62594105 on Build GRCh38
rs770442942 Chr.11: 62593995-62593995 on Build GRCh38
rs1046622373 Chr.11: 62593939-62593939 on Build GRCh38
rs753841609 Chr.11: 62593964-62593964 on Build GRCh38
rs754916174 Chr.11: 62593965-62593965 on Build GRCh38
rs777136719 Chr.11: 62593990-62593990 on Build GRCh38
rs540705606 Chr.11: 62593959-62593959 on Build GRCh38
rs200825288 Chr.11: 62593960-62593960 on Build GRCh38
rs751123747 Chr.11: 62593945-62593945 on Build GRCh38
rs772797368 Chr.11: 62594005-62594005 on Build GRCh38
rs758460310 Chr.11: 62594082-62594082 on Build GRCh38
rs773581090 Chr.11: 62593915-62593915 on Build GRCh38
rs767174807 Chr.11: 62594003-62594003 on Build GRCh38
rs747379658 Chr.11: 62593984-62593984 on Build GRCh38
rs761475332 Chr.11: 62593946-62593946 on Build GRCh38
rs752794500 Chr.11: 62594073-62594073 on Build GRCh38
rs748335784 Chr.11: 62593900-62593900 on Build GRCh38
rs748199521 Chr.11: 62593976-62593976 on Build GRCh38
rs778044780 Chr.11: 62594087-62594087 on Build GRCh38
rs142808404 Chr.11: 62594000-62594000 on Build GRCh38
rs1054336426 Chr.11: 62593951-62593951 on Build GRCh38
rs375087008 Chr.11: 62594089-62594089 on Build GRCh38
rs765194814 Chr.11: 62594037-62594037 on Build GRCh38
rs778907921 Chr.11: 62593970-62593970 on Build GRCh38
rs760059746 Chr.11: 62593994-62593994 on Build GRCh38
rs767107439 Chr.11: 62593952-62593952 on Build GRCh38
rs753647576 Chr.11: 62594027-62594027 on Build GRCh38
rs1019546612 Chr.11: 62594053-62594053 on Build GRCh38
rs573072156 Chr.11: 62593961-62593961 on Build GRCh38
rs771536567 Chr.11: 62593927-62593927 on Build GRCh38
rs771483181 Chr.11: 62593985-62593985 on Build GRCh38
rs769275642 Chr.11: 62593922-62593922 on Build GRCh38
rs112114991 Chr.11: 62594069-62594069 on Build GRCh38
rs775150148 Chr.11: 62593929-62593929 on Build GRCh38
rs760979939 Chr.11: 62593920-62593920 on Build GRCh38
rs371197494 Chr.11: 62594002-62594002 on Build GRCh38
rs777330447 Chr.11: 62593928-62593928 on Build GRCh38
rs917829378 Chr.11: 62594078-62594078 on Build GRCh38
rs778548131 Chr.11: 62593943-62593943 on Build GRCh38
rs994027467 Chr.11: 62594099-62594099 on Build GRCh38
rs754861081 Chr.11: 62594029-62594029 on Build GRCh38
rs1008868253 Chr.11: 62594036-62594036 on Build GRCh38
rs111400760 Chr.11: 62594006-62594006 on Build GRCh38

View Full Product Details