Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95049654-95049879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAL
Entrez Gene ID
4118
Gene Name
mal, T-cell differentiation protein
Gene Aliases
-
UniGene
Hs.80395
Gene Chromosome Location
Chr.2: 95025655-95053992 on Build GRCh38
Transcripts
NM_002371.3 NM_022438.2 NM_022439.2 NM_022440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4490692 c.367C>A p.H123N Chr.2: 95049686-95049686 on Build GRCh38
6446648 c.385G>A p.V129M Chr.2: 95049704-95049704 on Build GRCh38
4545866 c.387G>T p.V129V Chr.2: 95049706-95049706 on Build GRCh38
4648625 c.349T>C p.F117L Chr.2: 95049668-95049668 on Build GRCh38
3695639 c.359G>T p.R120M Chr.2: 95049678-95049678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531987339 Chr.2: 95049783-95049783 on Build GRCh38
rs983912073 Chr.2: 95049860-95049860 on Build GRCh38
rs965728479 Chr.2: 95049789-95049789 on Build GRCh38
rs372222607 Chr.2: 95049749-95049749 on Build GRCh38
rs776572875 Chr.2: 95049699-95049699 on Build GRCh38
rs756272728 Chr.2: 95049731-95049731 on Build GRCh38
rs772914357 Chr.2: 95049679-95049679 on Build GRCh38
rs750589227 Chr.2: 95049724-95049724 on Build GRCh38
rs370317076 Chr.2: 95049723-95049723 on Build GRCh38
rs770370071 Chr.2: 95049696-95049696 on Build GRCh38
rs769681793 Chr.2: 95049753-95049753 on Build GRCh38
rs546374228 Chr.2: 95049735-95049735 on Build GRCh38
rs367966598 Chr.2: 95049744-95049744 on Build GRCh38
rs1020961619 Chr.2: 95049781-95049781 on Build GRCh38
rs759026833 Chr.2: 95049752-95049752 on Build GRCh38
rs149416032 Chr.2: 95049703-95049703 on Build GRCh38
rs570607569 Chr.2: 95049743-95049743 on Build GRCh38
rs189368924 Chr.2: 95049809-95049809 on Build GRCh38
rs953911565 Chr.2: 95049829-95049829 on Build GRCh38
rs748518384 Chr.2: 95049742-95049742 on Build GRCh38
rs754426713 Chr.2: 95049739-95049739 on Build GRCh38
rs748648578 Chr.2: 95049841-95049841 on Build GRCh38
rs760185446 Chr.2: 95049685-95049685 on Build GRCh38
rs375041224 Chr.2: 95049740-95049740 on Build GRCh38
rs1028348103 Chr.2: 95049815-95049815 on Build GRCh38
rs762798081 Chr.2: 95049720-95049720 on Build GRCh38
rs201691542 Chr.2: 95049700-95049700 on Build GRCh38
rs776162591 Chr.2: 95049750-95049750 on Build GRCh38
rs138931615 Chr.2: 95049673-95049673 on Build GRCh38
rs112032328 Chr.2: 95049736-95049736 on Build GRCh38
rs754054137 Chr.2: 95049715-95049715 on Build GRCh38

View Full Product Details