Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30236263-30236508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB3
Entrez Gene ID
4114
Gene Name
MAGE family member B3
Gene Aliases
CT3.5
UniGene
Hs.113290
Gene Chromosome Location
Chr.X: 30230436-30237493 on Build GRCh38
Transcripts
NM_002365.4 XM_011545513.2 XM_011545514.2

SNP Detail

SNP ID Chromosome Location
rs778408289 Chr.X: 30236452-30236452 on Build GRCh38
rs771663707 Chr.X: 30236321-30236321 on Build GRCh38
rs778080700 Chr.X: 30236298-30236298 on Build GRCh38
rs752834151 Chr.X: 30236415-30236415 on Build GRCh38
rs760122075 Chr.X: 30236354-30236354 on Build GRCh38
rs920953044 Chr.X: 30236404-30236404 on Build GRCh38
rs757687731 Chr.X: 30236465-30236465 on Build GRCh38
rs775011357 Chr.X: 30236322-30236322 on Build GRCh38
rs2071310 Chr.X: 30236413-30236413 on Build GRCh38
rs34054171 Chr.X: 30236384-30236384 on Build GRCh38
rs754266826 Chr.X: 30236458-30236458 on Build GRCh38
rs745495531 Chr.X: 30236307-30236307 on Build GRCh38
rs760599195 Chr.X: 30236428-30236431 on Build GRCh38
rs764921111 Chr.X: 30236375-30236375 on Build GRCh38
rs768698062 Chr.X: 30236357-30236357 on Build GRCh38
rs769655784 Chr.X: 30236434-30236434 on Build GRCh38
rs778666100 Chr.X: 30236302-30236302 on Build GRCh38
rs776429024 Chr.X: 30236361-30236361 on Build GRCh38
rs963135681 Chr.X: 30236311-30236311 on Build GRCh38
rs752787599 Chr.X: 30236290-30236290 on Build GRCh38
rs973572898 Chr.X: 30236315-30236315 on Build GRCh38
rs113117104 Chr.X: 30236462-30236462 on Build GRCh38
rs756645824 Chr.X: 30236294-30236294 on Build GRCh38
rs746191660 Chr.X: 30236488-30236488 on Build GRCh38
rs761710997 Chr.X: 30236373-30236373 on Build GRCh38
rs750198978 Chr.X: 30236396-30236396 on Build GRCh38
rs975049397 Chr.X: 30236384-30236384 on Build GRCh38
rs779272274 Chr.X: 30236467-30236467 on Build GRCh38
rs749717727 Chr.X: 30236300-30236300 on Build GRCh38
rs759872010 Chr.X: 30236410-30236410 on Build GRCh38
rs771192209 Chr.X: 30236301-30236301 on Build GRCh38

View Full Product Details