Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57407680-57407953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM38
Entrez Gene ID
55544
Gene Name
RNA binding motif protein 38
Gene Aliases
HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2
UniGene
Hs.236361
Gene Chromosome Location
Chr.20: 57391398-57409333 on Build GRCh38
Transcripts
NM_001291780.1 NM_017495.5 NM_183425.2 XM_005260446.3 XM_011528885.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
263280 c.675C>T p.F225F Chr.20: 57407801-57407801 on Build GRCh38
4099710 c.628G>A p.A210T Chr.20: 57407754-57407754 on Build GRCh38
6764023 c.663G>A p.A221A Chr.20: 57407789-57407789 on Build GRCh38
7247055 c.652G>T p.A218S Chr.20: 57407778-57407778 on Build GRCh38
6269013 c.717G>T p.Q239H Chr.20: 57407843-57407843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764303895 Chr.20: 57407766-57407766 on Build GRCh38
rs376442730 Chr.20: 57407816-57407816 on Build GRCh38
rs758466011 Chr.20: 57407887-57407887 on Build GRCh38
rs776727871 Chr.20: 57407812-57407812 on Build GRCh38
rs774859528 Chr.20: 57407703-57407703 on Build GRCh38
rs769402847 Chr.20: 57407740-57407740 on Build GRCh38
rs751320408 Chr.20: 57407778-57407778 on Build GRCh38
rs748770068 Chr.20: 57407794-57407794 on Build GRCh38
rs199953546 Chr.20: 57407702-57407702 on Build GRCh38
rs908832017 Chr.20: 57407862-57407862 on Build GRCh38
rs746215170 Chr.20: 57407800-57407800 on Build GRCh38
rs760851249 Chr.20: 57407708-57407708 on Build GRCh38
rs201066490 Chr.20: 57407757-57407757 on Build GRCh38
rs368322258 Chr.20: 57407733-57407733 on Build GRCh38
rs572196610 Chr.20: 57407705-57407705 on Build GRCh38
rs780806248 Chr.20: 57407786-57407786 on Build GRCh38
rs949641830 Chr.20: 57407922-57407922 on Build GRCh38
rs200561320 Chr.20: 57407863-57407863 on Build GRCh38
rs771524419 Chr.20: 57407796-57407796 on Build GRCh38
rs1065290 Chr.20: 57407761-57407761 on Build GRCh38
rs771577545 Chr.20: 57407754-57407754 on Build GRCh38
rs753871234 Chr.20: 57407768-57407768 on Build GRCh38
rs201875738 Chr.20: 57407716-57407716 on Build GRCh38
rs542773204 Chr.20: 57407711-57407711 on Build GRCh38
rs760010932 Chr.20: 57407755-57407755 on Build GRCh38
rs755742871 Chr.20: 57407851-57407851 on Build GRCh38
rs779603862 Chr.20: 57407797-57407797 on Build GRCh38
rs774327187 Chr.20: 57407824-57407824 on Build GRCh38
rs543954479 Chr.20: 57407789-57407789 on Build GRCh38
rs1047780022 Chr.20: 57407707-57407707 on Build GRCh38
rs574040093 Chr.20: 57407706-57407706 on Build GRCh38
rs752446724 Chr.20: 57407846-57407846 on Build GRCh38
rs16980970 Chr.20: 57407802-57407802 on Build GRCh38
rs762894456 Chr.20: 57407771-57407771 on Build GRCh38
rs143107197 Chr.20: 57407756-57407756 on Build GRCh38
rs1034159207 Chr.20: 57407735-57407735 on Build GRCh38
rs199521379 Chr.20: 57407719-57407719 on Build GRCh38
rs772624606 Chr.20: 57407801-57407801 on Build GRCh38
rs754813917 Chr.20: 57407782-57407782 on Build GRCh38
rs183105885 Chr.20: 57407930-57407930 on Build GRCh38
rs765064896 Chr.20: 57407710-57407710 on Build GRCh38
rs759319181 Chr.20: 57407825-57407825 on Build GRCh38
rs201744631 Chr.20: 57407787-57407787 on Build GRCh38
rs573133688 Chr.20: 57407749-57407749 on Build GRCh38
rs373297597 Chr.20: 57407819-57407819 on Build GRCh38
rs780859344 Chr.20: 57407732-57407732 on Build GRCh38
rs751373448 Chr.20: 57407720-57407720 on Build GRCh38
rs924401044 Chr.20: 57407923-57407923 on Build GRCh38
rs766330885 Chr.20: 57407777-57407777 on Build GRCh38
rs1052644003 Chr.20: 57407931-57407931 on Build GRCh38
rs968380620 Chr.20: 57407906-57407906 on Build GRCh38
rs749736153 Chr.20: 57407753-57407753 on Build GRCh38
rs1019628330 Chr.20: 57407790-57407790 on Build GRCh38
rs777812060 Chr.20: 57407918-57407918 on Build GRCh38
rs754580963 Chr.20: 57407727-57407727 on Build GRCh38
rs373492567 Chr.20: 57407831-57407831 on Build GRCh38
rs1035920400 Chr.20: 57407843-57407843 on Build GRCh38
rs369246420 Chr.20: 57407788-57407788 on Build GRCh38
rs1065289 Chr.20: 57407725-57407725 on Build GRCh38

View Full Product Details