Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247965904-247966171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2AK2
Entrez Gene ID
391191
Gene Name
olfactory receptor family 2 subfamily AK member 2
Gene Aliases
OR1-47, OR2AK1P
UniGene
Hs.690413
Gene Chromosome Location
Chr.1: 247965332-247966339 on Build GRCh38
Transcripts
NM_001004491.1 NM_001304535.1 NM_175911.3

Gene Symbol
OR2L13
Entrez Gene ID
284521
Gene Name
olfactory receptor family 2 subfamily L member 13
Gene Aliases
OR2L14
UniGene
Hs.372936
Gene Chromosome Location
Chr.1: 247937029-248100922 on Build GRCh38
Transcripts
NM_001004491.1 NM_001304535.1 NM_175911.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2232790 c.791G>A p.S264N Chr.1: 247966122-247966122 on Build GRCh38
5821745 c.596T>A p.L199Q Chr.1: 247965927-247965927 on Build GRCh38
1241526 c.585A>C p.E195D Chr.1: 247965916-247965916 on Build GRCh38
7268900 c.771C>A p.S257S Chr.1: 247966102-247966102 on Build GRCh38
5705375 c.656T>C p.L219P Chr.1: 247965987-247965987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200437279 Chr.1: 247966075-247966075 on Build GRCh38
rs369499781 Chr.1: 247966111-247966111 on Build GRCh38
rs563425908 Chr.1: 247966069-247966069 on Build GRCh38
rs760387323 Chr.1: 247966038-247966038 on Build GRCh38
rs150750035 Chr.1: 247966146-247966146 on Build GRCh38
rs148880648 Chr.1: 247965949-247965949 on Build GRCh38
rs1008774565 Chr.1: 247965970-247965970 on Build GRCh38
rs752019788 Chr.1: 247966094-247966094 on Build GRCh38
rs75127345 Chr.1: 247965934-247965934 on Build GRCh38
rs748288102 Chr.1: 247966020-247966020 on Build GRCh38
rs750044627 Chr.1: 247965958-247965958 on Build GRCh38
rs371051095 Chr.1: 247966035-247966035 on Build GRCh38
rs1057059738 Chr.1: 247965987-247965987 on Build GRCh38
rs778494288 Chr.1: 247966007-247966007 on Build GRCh38
rs771003942 Chr.1: 247966029-247966029 on Build GRCh38
rs756642885 Chr.1: 247966116-247966116 on Build GRCh38
rs867145820 Chr.1: 247966092-247966092 on Build GRCh38
rs768691949 Chr.1: 247966048-247966048 on Build GRCh38
rs974352953 Chr.1: 247965953-247965953 on Build GRCh38
rs757222864 Chr.1: 247966107-247966107 on Build GRCh38
rs930148276 Chr.1: 247965956-247965956 on Build GRCh38
rs1044718224 Chr.1: 247966033-247966033 on Build GRCh38
rs141137318 Chr.1: 247966023-247966023 on Build GRCh38
rs140143594 Chr.1: 247966006-247966006 on Build GRCh38
rs761295580 Chr.1: 247965951-247965951 on Build GRCh38
rs753290116 Chr.1: 247966114-247966114 on Build GRCh38
rs761350666 Chr.1: 247966064-247966064 on Build GRCh38
rs4478844 Chr.1: 247965938-247965938 on Build GRCh38
rs778984527 Chr.1: 247966138-247966138 on Build GRCh38
rs755512639 Chr.1: 247966008-247966008 on Build GRCh38
rs753408812 Chr.1: 247965950-247965950 on Build GRCh38
rs369211584 Chr.1: 247966114-247966115 on Build GRCh38
rs767568518 Chr.1: 247966137-247966137 on Build GRCh38
rs548462465 Chr.1: 247966139-247966139 on Build GRCh38
rs891388584 Chr.1: 247965965-247965965 on Build GRCh38
rs749316861 Chr.1: 247966136-247966136 on Build GRCh38
rs765879946 Chr.1: 247966083-247966083 on Build GRCh38
rs200166151 Chr.1: 247966104-247966104 on Build GRCh38
rs74153219 Chr.1: 247966000-247966000 on Build GRCh38
rs777962249 Chr.1: 247966022-247966022 on Build GRCh38
rs574711780 Chr.1: 247966034-247966034 on Build GRCh38
rs769965428 Chr.1: 247966021-247966021 on Build GRCh38
rs764915853 Chr.1: 247965954-247965954 on Build GRCh38
rs762679392 Chr.1: 247966076-247966076 on Build GRCh38
rs903524641 Chr.1: 247966043-247966043 on Build GRCh38
rs143777155 Chr.1: 247966002-247966002 on Build GRCh38
rs776524052 Chr.1: 247966059-247966059 on Build GRCh38
rs778016379 Chr.1: 247966132-247966132 on Build GRCh38
rs866716443 Chr.1: 247965992-247965992 on Build GRCh38
rs764756091 Chr.1: 247966068-247966068 on Build GRCh38
rs781537668 Chr.1: 247966014-247966014 on Build GRCh38
rs756788410 Chr.1: 247965987-247965989 on Build GRCh38
rs1030930615 Chr.1: 247966124-247966124 on Build GRCh38

View Full Product Details