Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35563127-35563400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCDN
Entrez Gene ID
23154
Gene Name
neurochondrin
Gene Aliases
-
UniGene
Hs.121870
Gene Chromosome Location
Chr.1: 35557792-35566779 on Build GRCh38
Transcripts
NM_001014839.1 NM_001014841.1 NM_014284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007375 c.1417G>A p.V473I Chr.1: 35563233-35563233 on Build GRCh38
4007377 c.1430C>A p.P477H Chr.1: 35563246-35563246 on Build GRCh38
4705857 c.1355delG p.C452fs*3 Chr.1: 35563222-35563222 on Build GRCh38
4007372 c.1416C>T p.T472T Chr.1: 35563232-35563232 on Build GRCh38
2080580 c.1502C>T p.T501I Chr.1: 35563318-35563318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527931085 Chr.1: 35563164-35563164 on Build GRCh38
rs115546115 Chr.1: 35563211-35563211 on Build GRCh38
rs150386507 Chr.1: 35563331-35563331 on Build GRCh38
rs748812666 Chr.1: 35563246-35563246 on Build GRCh38
rs752035725 Chr.1: 35563322-35563322 on Build GRCh38
rs966846201 Chr.1: 35563245-35563245 on Build GRCh38
rs374733075 Chr.1: 35563248-35563248 on Build GRCh38
rs762010739 Chr.1: 35563279-35563279 on Build GRCh38
rs970858936 Chr.1: 35563159-35563159 on Build GRCh38
rs531256783 Chr.1: 35563286-35563286 on Build GRCh38
rs747807124 Chr.1: 35563226-35563226 on Build GRCh38
rs758052151 Chr.1: 35563168-35563168 on Build GRCh38
rs761818857 Chr.1: 35563178-35563178 on Build GRCh38
rs956053377 Chr.1: 35563233-35563233 on Build GRCh38
rs776779437 Chr.1: 35563277-35563277 on Build GRCh38
rs780949774 Chr.1: 35563216-35563216 on Build GRCh38
rs771863772 Chr.1: 35563271-35563271 on Build GRCh38
rs149457796 Chr.1: 35563359-35563359 on Build GRCh38
rs768006525 Chr.1: 35563332-35563332 on Build GRCh38
rs745639355 Chr.1: 35563269-35563269 on Build GRCh38
rs774201737 Chr.1: 35563254-35563254 on Build GRCh38
rs373852292 Chr.1: 35563232-35563232 on Build GRCh38
rs750560150 Chr.1: 35563186-35563186 on Build GRCh38
rs201630579 Chr.1: 35563313-35563313 on Build GRCh38
rs753167349 Chr.1: 35563339-35563339 on Build GRCh38
rs200776152 Chr.1: 35563368-35563368 on Build GRCh38
rs745810704 Chr.1: 35563366-35563366 on Build GRCh38
rs959370734 Chr.1: 35563179-35563179 on Build GRCh38
rs368069948 Chr.1: 35563355-35563355 on Build GRCh38
rs776871990 Chr.1: 35563169-35563169 on Build GRCh38
rs538755303 Chr.1: 35563363-35563363 on Build GRCh38
rs766859600 Chr.1: 35563320-35563320 on Build GRCh38
rs571289950 Chr.1: 35563356-35563356 on Build GRCh38
rs374688556 Chr.1: 35563185-35563185 on Build GRCh38
rs55707525 Chr.1: 35563190-35563190 on Build GRCh38
rs773247590 Chr.1: 35563292-35563292 on Build GRCh38
rs766837732 Chr.1: 35563196-35563196 on Build GRCh38
rs755819723 Chr.1: 35563346-35563346 on Build GRCh38

View Full Product Details