Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39315644-39315889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9
Entrez Gene ID
735
Gene Name
complement component 9
Gene Aliases
ARMD15, C9D
UniGene
Hs.654443
Gene Chromosome Location
Chr.5: 39284275-39364553 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2689712 c.812G>A p.R271Q Chr.5: 39315833-39315833 on Build GRCh38
6104177 c.798C>A p.G266G Chr.5: 39315847-39315847 on Build GRCh38
3616377 c.832G>A p.E278K Chr.5: 39315813-39315813 on Build GRCh38
6470362 c.860C>T p.S287F Chr.5: 39315785-39315785 on Build GRCh38
3616378 c.780C>T p.S260S Chr.5: 39315865-39315865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992587323 Chr.5: 39315798-39315798 on Build GRCh38
rs1057201528 Chr.5: 39315737-39315737 on Build GRCh38
rs188115836 Chr.5: 39315862-39315862 on Build GRCh38
rs1021420111 Chr.5: 39315717-39315718 on Build GRCh38
rs775577722 Chr.5: 39315861-39315861 on Build GRCh38
rs200524188 Chr.5: 39315833-39315833 on Build GRCh38
rs1011416686 Chr.5: 39315700-39315700 on Build GRCh38
rs974283690 Chr.5: 39315818-39315818 on Build GRCh38
rs137953033 Chr.5: 39315715-39315716 on Build GRCh38
rs757060107 Chr.5: 39315834-39315834 on Build GRCh38
rs775524568 Chr.5: 39315762-39315762 on Build GRCh38
rs745839437 Chr.5: 39315839-39315839 on Build GRCh38
rs766587251 Chr.5: 39315766-39315766 on Build GRCh38
rs1046874570 Chr.5: 39315854-39315854 on Build GRCh38
rs757468065 Chr.5: 39315699-39315699 on Build GRCh38
rs769722704 Chr.5: 39315840-39315840 on Build GRCh38
rs756959099 Chr.5: 39315850-39315850 on Build GRCh38
rs776375864 Chr.5: 39315746-39315746 on Build GRCh38
rs899187184 Chr.5: 39315777-39315777 on Build GRCh38
rs777500599 Chr.5: 39315821-39315821 on Build GRCh38
rs867203884 Chr.5: 39315813-39315813 on Build GRCh38
rs764259892 Chr.5: 39315779-39315779 on Build GRCh38
rs934855573 Chr.5: 39315701-39315701 on Build GRCh38
rs191850540 Chr.5: 39315787-39315787 on Build GRCh38
rs374455227 Chr.5: 39315719-39315720 on Build GRCh38
rs764893805 Chr.5: 39315806-39315806 on Build GRCh38
rs746761191 Chr.5: 39315739-39315739 on Build GRCh38
rs778737314 Chr.5: 39315807-39315807 on Build GRCh38
rs766483572 Chr.5: 39315842-39315842 on Build GRCh38
rs370941399 Chr.5: 39315748-39315748 on Build GRCh38
rs948817955 Chr.5: 39315756-39315756 on Build GRCh38
rs767908678 Chr.5: 39315800-39315800 on Build GRCh38
rs542090983 Chr.5: 39315697-39315697 on Build GRCh38
rs149201228 Chr.5: 39315795-39315795 on Build GRCh38
rs770906281 Chr.5: 39315741-39315741 on Build GRCh38
rs763026372 Chr.5: 39315782-39315787 on Build GRCh38
rs781007927 Chr.5: 39315835-39315835 on Build GRCh38
rs756616544 Chr.5: 39315801-39315801 on Build GRCh38
rs183125896 Chr.5: 39315810-39315810 on Build GRCh38
rs769803219 Chr.5: 39315751-39315751 on Build GRCh38
rs34625111 Chr.5: 39315809-39315809 on Build GRCh38
rs368138699 Chr.5: 39315778-39315778 on Build GRCh38
rs1044402484 Chr.5: 39315775-39315775 on Build GRCh38
rs995136799 Chr.5: 39315785-39315785 on Build GRCh38

View Full Product Details