Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143478155-143478428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA1-AS1
Entrez Gene ID
285965
Gene Name
EPHA1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.642649
Gene Chromosome Location
Chr.7: 143407813-143523449 on Build GRCh38
Transcripts
NR_033897.1 NM_176883.2

Gene Symbol
TAS2R41
Entrez Gene ID
259287
Gene Name
taste 2 receptor member 41
Gene Aliases
T2R41, T2R59
UniGene
Hs.650648
Gene Chromosome Location
Chr.7: 143477873-143478796 on Build GRCh38
Transcripts
NR_033897.1 NM_176883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3258393 c.516G>A p.W172* Chr.7: 143478388-143478388 on Build GRCh38
6797004 c.368A>G p.K123R Chr.7: 143478240-143478240 on Build GRCh38
375893 c.427A>T p.I143F Chr.7: 143478299-143478299 on Build GRCh38
3258391 c.442delT p.W150fs*2 Chr.7: 143478314-143478314 on Build GRCh38
3745596 c.347A>G p.H116R Chr.7: 143478219-143478219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200798577 Chr.7: 143478285-143478285 on Build GRCh38
rs552745650 Chr.7: 143478295-143478295 on Build GRCh38
rs775041713 Chr.7: 143478204-143478204 on Build GRCh38
rs769289974 Chr.7: 143478190-143478190 on Build GRCh38
rs372486115 Chr.7: 143478288-143478288 on Build GRCh38
rs760748906 Chr.7: 143478247-143478247 on Build GRCh38
rs761694543 Chr.7: 143478223-143478223 on Build GRCh38
rs766448538 Chr.7: 143478320-143478320 on Build GRCh38
rs778808879 Chr.7: 143478310-143478310 on Build GRCh38
rs745318041 Chr.7: 143478182-143478182 on Build GRCh38
rs767457860 Chr.7: 143478378-143478378 on Build GRCh38
rs573900957 Chr.7: 143478299-143478299 on Build GRCh38
rs773979221 Chr.7: 143478221-143478221 on Build GRCh38
rs760656550 Chr.7: 143478313-143478313 on Build GRCh38
rs1005528657 Chr.7: 143478210-143478210 on Build GRCh38
rs897108982 Chr.7: 143478254-143478254 on Build GRCh38
rs534710446 Chr.7: 143478259-143478259 on Build GRCh38
rs781728761 Chr.7: 143478179-143478179 on Build GRCh38
rs373019967 Chr.7: 143478290-143478290 on Build GRCh38
rs912895230 Chr.7: 143478400-143478400 on Build GRCh38
rs754530036 Chr.7: 143478208-143478209 on Build GRCh38
rs763138122 Chr.7: 143478335-143478335 on Build GRCh38
rs756576784 Chr.7: 143478391-143478391 on Build GRCh38
rs747835202 Chr.7: 143478303-143478303 on Build GRCh38
rs538412437 Chr.7: 143478385-143478385 on Build GRCh38
rs751801832 Chr.7: 143478360-143478360 on Build GRCh38
rs373715039 Chr.7: 143478283-143478283 on Build GRCh38
rs571117123 Chr.7: 143478331-143478331 on Build GRCh38
rs771883355 Chr.7: 143478305-143478305 on Build GRCh38
rs753168244 Chr.7: 143478313-143478314 on Build GRCh38
rs377524079 Chr.7: 143478341-143478341 on Build GRCh38
rs748830538 Chr.7: 143478289-143478289 on Build GRCh38
rs773192241 Chr.7: 143478246-143478246 on Build GRCh38
rs770609445 Chr.7: 143478307-143478307 on Build GRCh38
rs764397844 Chr.7: 143478266-143478266 on Build GRCh38
rs766087090 Chr.7: 143478193-143478193 on Build GRCh38
rs146208812 Chr.7: 143478260-143478260 on Build GRCh38
rs369344852 Chr.7: 143478272-143478272 on Build GRCh38
rs767902819 Chr.7: 143478381-143478381 on Build GRCh38
rs10278721 Chr.7: 143478252-143478252 on Build GRCh38
rs753474078 Chr.7: 143478204-143478205 on Build GRCh38
rs1029540439 Chr.7: 143478279-143478279 on Build GRCh38
rs377473928 Chr.7: 143478284-143478284 on Build GRCh38
rs768341070 Chr.7: 143478218-143478218 on Build GRCh38
rs757480222 Chr.7: 143478363-143478363 on Build GRCh38
rs751812332 Chr.7: 143478271-143478271 on Build GRCh38
rs267601367 Chr.7: 143478343-143478343 on Build GRCh38
rs764688543 Chr.7: 143478232-143478246 on Build GRCh38
rs748918541 Chr.7: 143478209-143478209 on Build GRCh38

View Full Product Details