Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 159034494-159034767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPR2
Entrez Gene ID
7434
Gene Name
vasoactive intestinal peptide receptor 2
Gene Aliases
C16DUPq36.3, DUP7q36.3, PACAP-R-3, PACAP-R3, VIP-R-2, VPAC2, VPAC2R, VPCAP2R
UniGene
Hs.585052
Gene Chromosome Location
Chr.7: 159028175-159144957 on Build GRCh38
Transcripts
NM_001304522.1 NM_001308259.1 NM_003382.4 NR_130758.1 XM_005249561.3 XM_006716107.2 XM_006716108.3 XM_011516550.2 XM_017012580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762545 c.825C>T p.N275N Chr.7: 159034635-159034635 on Build GRCh38
4164241 c.879+1G>A p.? Chr.7: 159034580-159034580 on Build GRCh38
1088036 c.869T>G p.I290S Chr.7: 159034591-159034591 on Build GRCh38
1699678 c.854G>A p.R285Q Chr.7: 159034606-159034606 on Build GRCh38
3636882 c.879C>T p.I293I Chr.7: 159034581-159034581 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs970576727 Chr.7: 159034741-159034741 on Build GRCh38
rs753287663 Chr.7: 159034615-159034615 on Build GRCh38
rs538920701 Chr.7: 159034713-159034713 on Build GRCh38
rs774603165 Chr.7: 159034686-159034686 on Build GRCh38
rs750350963 Chr.7: 159034609-159034609 on Build GRCh38
rs182279904 Chr.7: 159034728-159034728 on Build GRCh38
rs564720397 Chr.7: 159034571-159034571 on Build GRCh38
rs778534104 Chr.7: 159034630-159034630 on Build GRCh38
rs928431825 Chr.7: 159034721-159034721 on Build GRCh38
rs188877591 Chr.7: 159034719-159034719 on Build GRCh38
rs746279223 Chr.7: 159034651-159034651 on Build GRCh38
rs2071624 Chr.7: 159034635-159034635 on Build GRCh38
rs775675627 Chr.7: 159034584-159034584 on Build GRCh38
rs775442068 Chr.7: 159034642-159034643 on Build GRCh38
rs199630455 Chr.7: 159034586-159034586 on Build GRCh38
rs778550594 Chr.7: 159034559-159034559 on Build GRCh38
rs763115583 Chr.7: 159034683-159034683 on Build GRCh38
rs559239908 Chr.7: 159034716-159034716 on Build GRCh38
rs551665698 Chr.7: 159034533-159034533 on Build GRCh38
rs936715335 Chr.7: 159034604-159034604 on Build GRCh38
rs762063586 Chr.7: 159034555-159034555 on Build GRCh38
rs770363078 Chr.7: 159034653-159034653 on Build GRCh38
rs2071623 Chr.7: 159034723-159034723 on Build GRCh38
rs913346696 Chr.7: 159034735-159034735 on Build GRCh38
rs750145139 Chr.7: 159034700-159034700 on Build GRCh38
rs755996366 Chr.7: 159034610-159034610 on Build GRCh38
rs763019054 Chr.7: 159034678-159034679 on Build GRCh38
rs777167573 Chr.7: 159034566-159034566 on Build GRCh38
rs779998027 Chr.7: 159034611-159034611 on Build GRCh38
rs947477344 Chr.7: 159034740-159034741 on Build GRCh38
rs143405207 Chr.7: 159034599-159034599 on Build GRCh38
rs781365999 Chr.7: 159034648-159034648 on Build GRCh38
rs748818486 Chr.7: 159034546-159034546 on Build GRCh38
rs776149910 Chr.7: 159034669-159034669 on Build GRCh38
rs771934064 Chr.7: 159034702-159034702 on Build GRCh38
rs886203911 Chr.7: 159034715-159034715 on Build GRCh38
rs150965113 Chr.7: 159034600-159034600 on Build GRCh38
rs542320002 Chr.7: 159034743-159034743 on Build GRCh38
rs767254624 Chr.7: 159034607-159034607 on Build GRCh38
rs376298219 Chr.7: 159034580-159034580 on Build GRCh38
rs189642032 Chr.7: 159034720-159034720 on Build GRCh38
rs142549613 Chr.7: 159034579-159034579 on Build GRCh38
rs370731634 Chr.7: 159034606-159034606 on Build GRCh38
rs1017033 Chr.7: 159034710-159034710 on Build GRCh38
rs184029542 Chr.7: 159034694-159034694 on Build GRCh38
rs879002122 Chr.7: 159034627-159034627 on Build GRCh38
rs771267900 Chr.7: 159034563-159034563 on Build GRCh38
rs746157412 Chr.7: 159034546-159034547 on Build GRCh38
rs959326419 Chr.7: 159034722-159034722 on Build GRCh38
rs41271229 Chr.7: 159034695-159034695 on Build GRCh38
rs997266704 Chr.7: 159034634-159034634 on Build GRCh38
rs770041606 Chr.7: 159034578-159034579 on Build GRCh38
rs768927585 Chr.7: 159034684-159034684 on Build GRCh38
rs571344640 Chr.7: 159034534-159034534 on Build GRCh38
rs747798339 Chr.7: 159034632-159034632 on Build GRCh38
rs991166657 Chr.7: 159034573-159034573 on Build GRCh38
rs754542029 Chr.7: 159034625-159034625 on Build GRCh38
rs143209252 Chr.7: 159034637-159034637 on Build GRCh38

View Full Product Details