Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136674940-136675165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF6
Entrez Gene ID
9459
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor 6
Gene Aliases
COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX
UniGene
Hs.522795
Gene Chromosome Location
Chr.X: 136665547-136781656 on Build GRCh38
Transcripts
NM_001306177.1 NM_004840.2 XM_005262499.2 XM_011531412.2 XM_011531413.1 XM_011531414.1 XM_011531415.2 XM_011531416.2 XM_011531417.1 XM_017029975.1 XM_017029976.1

SNP Detail

SNP ID Chromosome Location
rs765154485 Chr.X: 136675013-136675013 on Build GRCh38
rs188922132 Chr.X: 136675134-136675134 on Build GRCh38
rs761503931 Chr.X: 136675041-136675041 on Build GRCh38
rs760790388 Chr.X: 136675004-136675004 on Build GRCh38
rs970133862 Chr.X: 136675023-136675023 on Build GRCh38
rs775500896 Chr.X: 136675034-136675034 on Build GRCh38
rs773210061 Chr.X: 136675001-136675001 on Build GRCh38
rs769158910 Chr.X: 136674982-136674982 on Build GRCh38
rs767515682 Chr.X: 136675137-136675137 on Build GRCh38
rs757618931 Chr.X: 136674998-136674998 on Build GRCh38
rs755657383 Chr.X: 136675099-136675099 on Build GRCh38
rs749694794 Chr.X: 136674971-136674971 on Build GRCh38
rs376908686 Chr.X: 136675053-136675053 on Build GRCh38
rs928424896 Chr.X: 136675095-136675095 on Build GRCh38
rs779377174 Chr.X: 136675114-136675114 on Build GRCh38
rs749971894 Chr.X: 136675065-136675065 on Build GRCh38
rs1053351571 Chr.X: 136675133-136675133 on Build GRCh38
rs763897398 Chr.X: 136675037-136675037 on Build GRCh38
rs751326434 Chr.X: 136675039-136675039 on Build GRCh38
rs12008084 Chr.X: 136675035-136675035 on Build GRCh38
rs748410522 Chr.X: 136674996-136674996 on Build GRCh38
rs774810336 Chr.X: 136674989-136674989 on Build GRCh38

View Full Product Details