Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48134685-48134948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYEF2
Entrez Gene ID
50804
Gene Name
myelin expression factor 2
Gene Aliases
HsT18564, MEF-2, MST156, MSTP156, myEF-2
UniGene
Hs.653123
Gene Chromosome Location
Chr.15: 48134632-48178517 on Build GRCh38
Transcripts
XM_005254425.4 XM_017022285.1 XM_017022286.1 XM_017022287.1 XM_017022291.1 XM_017022292.1 NM_205850.2 XM_017022079.1 XM_017022080.1

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
UniGene
Hs.710240
Gene Chromosome Location
Chr.15: 48120972-48142392 on Build GRCh38
Transcripts
XM_005254425.4 XM_017022285.1 XM_017022286.1 XM_017022287.1 XM_017022291.1 XM_017022292.1 NM_205850.2 XM_017022079.1 XM_017022080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
145940 c.541A>G p.I181V Chr.15: 48134935-48134935 on Build GRCh38
2189943 c.534G>A p.A178A Chr.15: 48134928-48134928 on Build GRCh38
962309 c.490-1G>T p.? Chr.15: 48134883-48134883 on Build GRCh38
5821280 c.520A>T p.R174* Chr.15: 48134914-48134914 on Build GRCh38
6077397 c.533C>G p.A178G Chr.15: 48134927-48134927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757159427 Chr.15: 48134856-48134856 on Build GRCh38
rs556168142 Chr.15: 48134927-48134927 on Build GRCh38
rs374448307 Chr.15: 48134899-48134899 on Build GRCh38
rs577617254 Chr.15: 48134779-48134779 on Build GRCh38
rs762693511 Chr.15: 48134847-48134847 on Build GRCh38
rs1013978461 Chr.15: 48134852-48134852 on Build GRCh38
rs183612933 Chr.15: 48134819-48134819 on Build GRCh38
rs977118193 Chr.15: 48134897-48134897 on Build GRCh38
rs752641969 Chr.15: 48134896-48134896 on Build GRCh38
rs897817691 Chr.15: 48134742-48134742 on Build GRCh38
rs770538169 Chr.15: 48134872-48134872 on Build GRCh38
rs767431083 Chr.15: 48134892-48134892 on Build GRCh38
rs958807671 Chr.15: 48134849-48134849 on Build GRCh38
rs1008996682 Chr.15: 48134783-48134783 on Build GRCh38
rs774164778 Chr.15: 48134881-48134881 on Build GRCh38
rs964899264 Chr.15: 48134807-48134807 on Build GRCh38
rs1046964367 Chr.15: 48134805-48134805 on Build GRCh38
rs957604752 Chr.15: 48134908-48134908 on Build GRCh38
rs759318202 Chr.15: 48134891-48134891 on Build GRCh38
rs555994428 Chr.15: 48134907-48134907 on Build GRCh38
rs904191326 Chr.15: 48134744-48134744 on Build GRCh38
rs1052865486 Chr.15: 48134751-48134752 on Build GRCh38
rs113104600 Chr.15: 48134751-48134751 on Build GRCh38
rs1000308736 Chr.15: 48134750-48134750 on Build GRCh38
rs771026148 Chr.15: 48134908-48134909 on Build GRCh38
rs772962339 Chr.15: 48134845-48134845 on Build GRCh38
rs867245480 Chr.15: 48134816-48134816 on Build GRCh38
rs769436678 Chr.15: 48134837-48134837 on Build GRCh38
rs749313900 Chr.15: 48134890-48134897 on Build GRCh38
rs891563835 Chr.15: 48134767-48134767 on Build GRCh38

View Full Product Details