Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22554410-22554671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORBS3
Entrez Gene ID
10174
Gene Name
sorbin and SH3 domain containing 3
Gene Aliases
SCAM-1, SCAM1, SH3D4
UniGene
Hs.528572
Gene Chromosome Location
Chr.8: 22544990-22575495 on Build GRCh38
Transcripts
NM_005775.4 XM_006716266.1 XM_006716268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5598806 c.11C>T p.P4L Chr.8: 22554517-22554517 on Build GRCh38
2786192 c.27C>T p.R9R Chr.8: 22554533-22554533 on Build GRCh38
6435409 c.23T>A p.L8H Chr.8: 22554529-22554529 on Build GRCh38
4500220 c.55C>T p.P19S Chr.8: 22554561-22554561 on Build GRCh38
1456099 c.46G>A p.D16N Chr.8: 22554552-22554552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529403457 Chr.8: 22554632-22554632 on Build GRCh38
rs753866090 Chr.8: 22554589-22554589 on Build GRCh38
rs752370439 Chr.8: 22554600-22554600 on Build GRCh38
rs370407437 Chr.8: 22554598-22554598 on Build GRCh38
rs887913918 Chr.8: 22554641-22554641 on Build GRCh38
rs934487997 Chr.8: 22554594-22554595 on Build GRCh38
rs748833925 Chr.8: 22554553-22554553 on Build GRCh38
rs201796577 Chr.8: 22554567-22554567 on Build GRCh38
rs751918846 Chr.8: 22554647-22554647 on Build GRCh38
rs377013679 Chr.8: 22554633-22554633 on Build GRCh38
rs1027923819 Chr.8: 22554635-22554635 on Build GRCh38
rs774264751 Chr.8: 22554515-22554515 on Build GRCh38
rs547500889 Chr.8: 22554648-22554648 on Build GRCh38
rs372965024 Chr.8: 22554524-22554524 on Build GRCh38
rs138529127 Chr.8: 22554522-22554522 on Build GRCh38
rs766945481 Chr.8: 22554645-22554645 on Build GRCh38
rs146179804 Chr.8: 22554532-22554532 on Build GRCh38
rs867588341 Chr.8: 22554555-22554555 on Build GRCh38
rs998746132 Chr.8: 22554431-22554431 on Build GRCh38
rs149634131 Chr.8: 22554578-22554578 on Build GRCh38
rs760494255 Chr.8: 22554621-22554621 on Build GRCh38
rs1035679715 Chr.8: 22554447-22554447 on Build GRCh38
rs750961338 Chr.8: 22554597-22554597 on Build GRCh38
rs574478872 Chr.8: 22554429-22554432 on Build GRCh38
rs760935746 Chr.8: 22554576-22554576 on Build GRCh38
rs748745989 Chr.8: 22554602-22554602 on Build GRCh38
rs1042080297 Chr.8: 22554594-22554594 on Build GRCh38
rs576640305 Chr.8: 22554459-22554459 on Build GRCh38
rs775188135 Chr.8: 22554609-22554609 on Build GRCh38
rs907968226 Chr.8: 22554521-22554521 on Build GRCh38
rs139783217 Chr.8: 22554549-22554549 on Build GRCh38
rs747630111 Chr.8: 22554481-22554481 on Build GRCh38
rs771827568 Chr.8: 22554568-22554568 on Build GRCh38
rs983508520 Chr.8: 22554517-22554517 on Build GRCh38
rs747870516 Chr.8: 22554548-22554548 on Build GRCh38
rs765709663 Chr.8: 22554590-22554590 on Build GRCh38
rs144354172 Chr.8: 22554601-22554601 on Build GRCh38
rs35511419 Chr.8: 22554608-22554608 on Build GRCh38
rs181973729 Chr.8: 22554607-22554607 on Build GRCh38
rs148763733 Chr.8: 22554606-22554606 on Build GRCh38
rs761650588 Chr.8: 22554516-22554541 on Build GRCh38
rs966181093 Chr.8: 22554649-22554649 on Build GRCh38
rs558503978 Chr.8: 22554458-22554458 on Build GRCh38
rs994517542 Chr.8: 22554615-22554615 on Build GRCh38
rs546463155 Chr.8: 22554573-22554573 on Build GRCh38
rs753608739 Chr.8: 22554651-22554651 on Build GRCh38
rs752863734 Chr.8: 22554526-22554526 on Build GRCh38
rs373300127 Chr.8: 22554556-22554556 on Build GRCh38
rs780895468 Chr.8: 22554545-22554545 on Build GRCh38
rs765293608 Chr.8: 22554637-22554637 on Build GRCh38
rs141486783 Chr.8: 22554531-22554531 on Build GRCh38
rs766250044 Chr.8: 22554490-22554490 on Build GRCh38
rs759267569 Chr.8: 22554513-22554513 on Build GRCh38
rs771785495 Chr.8: 22554519-22554519 on Build GRCh38
rs376000430 Chr.8: 22554629-22554629 on Build GRCh38
rs779309968 Chr.8: 22554533-22554533 on Build GRCh38
rs370177041 Chr.8: 22554482-22554482 on Build GRCh38
rs774152106 Chr.8: 22554495-22554495 on Build GRCh38
rs189601642 Chr.8: 22554484-22554484 on Build GRCh38
rs756180273 Chr.8: 22554530-22554530 on Build GRCh38
rs369770694 Chr.8: 22554551-22554551 on Build GRCh38
rs111422443 Chr.8: 22554487-22554487 on Build GRCh38
rs1003212268 Chr.8: 22554435-22554435 on Build GRCh38
rs763413227 Chr.8: 22554644-22554644 on Build GRCh38
rs774166838 Chr.8: 22554562-22554562 on Build GRCh38
rs750923529 Chr.8: 22554642-22554642 on Build GRCh38
rs112969836 Chr.8: 22554534-22554534 on Build GRCh38
rs1043179484 Chr.8: 22554552-22554552 on Build GRCh38

View Full Product Details