Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40323911-40324182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
UniGene
Hs.631557
Gene Chromosome Location
Chr.19: 40292743-40348527 on Build GRCh38
Transcripts
NM_001256440.1 NM_001256441.1 NR_046202.1 XM_005258520.2 XM_011526460.2 XM_017026291.1 XM_017026292.1 XM_017026293.1 XM_017026294.1 XM_017026295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078168 c.573+1G>T p.? Chr.19: 40324005-40324005 on Build GRCh38
996636 c.550G>T p.A184S Chr.19: 40324029-40324029 on Build GRCh38
474745 c.536G>A p.G179D Chr.19: 40324043-40324043 on Build GRCh38
4078170 c.515C>T p.T172M Chr.19: 40324064-40324064 on Build GRCh38
996637 c.507C>A p.L169L Chr.19: 40324072-40324072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568525596 Chr.19: 40323987-40323987 on Build GRCh38
rs972074756 Chr.19: 40324041-40324041 on Build GRCh38
rs765454585 Chr.19: 40323996-40323996 on Build GRCh38
rs771813525 Chr.19: 40324023-40324023 on Build GRCh38
rs770369356 Chr.19: 40323977-40323990 on Build GRCh38
rs904589089 Chr.19: 40323934-40323934 on Build GRCh38
rs757537235 Chr.19: 40323993-40323993 on Build GRCh38
rs781765898 Chr.19: 40324085-40324085 on Build GRCh38
rs890357737 Chr.19: 40324137-40324137 on Build GRCh38
rs766693508 Chr.19: 40324064-40324064 on Build GRCh38
rs776165874 Chr.19: 40324108-40324108 on Build GRCh38
rs745804894 Chr.19: 40323985-40323985 on Build GRCh38
rs747650981 Chr.19: 40323965-40323965 on Build GRCh38
rs1008883316 Chr.19: 40323957-40323957 on Build GRCh38
rs1053607458 Chr.19: 40324139-40324139 on Build GRCh38
rs1014288338 Chr.19: 40323976-40323976 on Build GRCh38
rs756851403 Chr.19: 40324097-40324097 on Build GRCh38
rs774230071 Chr.19: 40323975-40323975 on Build GRCh38
rs375387543 Chr.19: 40324042-40324042 on Build GRCh38
rs201282159 Chr.19: 40323992-40323992 on Build GRCh38
rs370778260 Chr.19: 40323945-40323945 on Build GRCh38
rs764224295 Chr.19: 40324039-40324039 on Build GRCh38
rs772190895 Chr.19: 40324116-40324116 on Build GRCh38
rs760788312 Chr.19: 40323986-40323986 on Build GRCh38
rs891915786 Chr.19: 40324144-40324144 on Build GRCh38
rs775470099 Chr.19: 40324024-40324024 on Build GRCh38
rs372052708 Chr.19: 40324015-40324015 on Build GRCh38
rs543163328 Chr.19: 40323971-40323971 on Build GRCh38
rs767450526 Chr.19: 40323980-40323980 on Build GRCh38
rs376546479 Chr.19: 40324106-40324106 on Build GRCh38
rs775313438 Chr.19: 40323981-40323981 on Build GRCh38
rs748768374 Chr.19: 40324090-40324090 on Build GRCh38
rs772896869 Chr.19: 40323967-40323967 on Build GRCh38
rs201580687 Chr.19: 40324081-40324081 on Build GRCh38
rs572467559 Chr.19: 40323950-40323950 on Build GRCh38
rs755618274 Chr.19: 40324018-40324018 on Build GRCh38
rs867200152 Chr.19: 40324118-40324118 on Build GRCh38
rs769207411 Chr.19: 40323966-40323966 on Build GRCh38
rs200609412 Chr.19: 40324021-40324021 on Build GRCh38
rs368410723 Chr.19: 40324103-40324103 on Build GRCh38
rs150788237 Chr.19: 40324044-40324044 on Build GRCh38
rs745611374 Chr.19: 40324022-40324022 on Build GRCh38
rs771899789 Chr.19: 40324110-40324110 on Build GRCh38
rs1039206982 Chr.19: 40323931-40323931 on Build GRCh38
rs758717726 Chr.19: 40324012-40324012 on Build GRCh38
rs747517123 Chr.19: 40324013-40324013 on Build GRCh38
rs746839091 Chr.19: 40324122-40324122 on Build GRCh38
rs551537904 Chr.19: 40324045-40324045 on Build GRCh38
rs770845444 Chr.19: 40323972-40323972 on Build GRCh38
rs750716786 Chr.19: 40324000-40324000 on Build GRCh38
rs750663694 Chr.19: 40324063-40324063 on Build GRCh38
rs371427678 Chr.19: 40324121-40324121 on Build GRCh38
rs760599992 Chr.19: 40324038-40324038 on Build GRCh38
rs139156915 Chr.19: 40324069-40324069 on Build GRCh38
rs199793069 Chr.19: 40324019-40324019 on Build GRCh38
rs761904752 Chr.19: 40324112-40324112 on Build GRCh38

View Full Product Details