Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2430949-2431190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
UniGene
Hs.1973
Gene Chromosome Location
Chr.16: 2425438-2458858 on Build GRCh38
Transcripts
NM_001323538.1 NM_001761.2 XM_017023822.1 XM_017023823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059507 c.62G>A p.R21Q Chr.16: 2431175-2431175 on Build GRCh38
5384573 c.53C>T p.P18L Chr.16: 2431166-2431166 on Build GRCh38
5757836 c.34G>A p.A12T Chr.16: 2431147-2431147 on Build GRCh38
6195103 c.45C>T p.F15F Chr.16: 2431158-2431158 on Build GRCh38
6078761 c.17-2A>T p.? Chr.16: 2431128-2431128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775895715 Chr.16: 2431054-2431054 on Build GRCh38
rs755670344 Chr.16: 2431088-2431088 on Build GRCh38
rs775549664 Chr.16: 2430995-2430995 on Build GRCh38
rs747010806 Chr.16: 2431113-2431113 on Build GRCh38
rs773521452 Chr.16: 2431151-2431151 on Build GRCh38
rs894690859 Chr.16: 2431002-2431002 on Build GRCh38
rs747062018 Chr.16: 2431000-2431000 on Build GRCh38
rs1014522700 Chr.16: 2431012-2431012 on Build GRCh38
rs748315822 Chr.16: 2431132-2431132 on Build GRCh38
rs375454117 Chr.16: 2431111-2431111 on Build GRCh38
rs745629009 Chr.16: 2431105-2431105 on Build GRCh38
rs142241270 Chr.16: 2431164-2431164 on Build GRCh38
rs763420643 Chr.16: 2431048-2431048 on Build GRCh38
rs768793571 Chr.16: 2431019-2431019 on Build GRCh38
rs531239350 Chr.16: 2431089-2431089 on Build GRCh38
rs533405080 Chr.16: 2431127-2431127 on Build GRCh38
rs762039769 Chr.16: 2431043-2431043 on Build GRCh38
rs752173112 Chr.16: 2431081-2431081 on Build GRCh38
rs747247974 Chr.16: 2431107-2431107 on Build GRCh38
rs765574299 Chr.16: 2431045-2431045 on Build GRCh38
rs753473829 Chr.16: 2431096-2431096 on Build GRCh38
rs957837689 Chr.16: 2431084-2431084 on Build GRCh38
rs772604305 Chr.16: 2431010-2431011 on Build GRCh38
rs367591984 Chr.16: 2431108-2431108 on Build GRCh38
rs764855503 Chr.16: 2431116-2431116 on Build GRCh38
rs369290461 Chr.16: 2430989-2430989 on Build GRCh38
rs745988003 Chr.16: 2431009-2431010 on Build GRCh38
rs761552584 Chr.16: 2431098-2431099 on Build GRCh38
rs375100764 Chr.16: 2430987-2430987 on Build GRCh38
rs776824044 Chr.16: 2431030-2431030 on Build GRCh38
rs979844154 Chr.16: 2431077-2431077 on Build GRCh38
rs9936014 Chr.16: 2431103-2431103 on Build GRCh38
rs982770088 Chr.16: 2431133-2431133 on Build GRCh38
rs757002386 Chr.16: 2431102-2431102 on Build GRCh38
rs764562957 Chr.16: 2431062-2431063 on Build GRCh38
rs548054377 Chr.16: 2431053-2431054 on Build GRCh38
rs766763090 Chr.16: 2431059-2431059 on Build GRCh38
rs769881023 Chr.16: 2431137-2431137 on Build GRCh38
rs776351633 Chr.16: 2431063-2431063 on Build GRCh38
rs111704404 Chr.16: 2431128-2431128 on Build GRCh38

View Full Product Details