Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27368239-27368464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL2
Entrez Gene ID
56938
Gene Name
aryl hydrocarbon receptor nuclear translocator like 2
Gene Aliases
BMAL2, CLIF, MOP9, PASD9, bHLHe6
UniGene
Hs.445447
Gene Chromosome Location
Chr.12: 27332854-27425813 on Build GRCh38
Transcripts
NM_001248002.1 NM_001248003.1 NM_001248004.1 NM_001248005.1 NM_020183.4 XM_006719112.3 XM_011520766.2 XM_011520768.2 XM_011520769.2 XM_011520770.2 XM_017019666.1 XM_017019667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
184384 c.148C>T p.R50* Chr.12: 27368378-27368378 on Build GRCh38
1283509 c.148C>A p.R50R Chr.12: 27368378-27368378 on Build GRCh38
1361070 c.83G>A p.R28H Chr.12: 27368313-27368313 on Build GRCh38
6605099 c.154C>T p.R52C Chr.12: 27368384-27368384 on Build GRCh38
3792477 c.44G>C p.R15T Chr.12: 27368274-27368274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764534769 Chr.12: 27368339-27368339 on Build GRCh38
rs756251882 Chr.12: 27368376-27368376 on Build GRCh38
rs140404198 Chr.12: 27368390-27368390 on Build GRCh38
rs776246315 Chr.12: 27368315-27368315 on Build GRCh38
rs749536664 Chr.12: 27368384-27368384 on Build GRCh38
rs185375677 Chr.12: 27368325-27368325 on Build GRCh38
rs140915764 Chr.12: 27368314-27368314 on Build GRCh38
rs774421945 Chr.12: 27368334-27368336 on Build GRCh38
rs771903769 Chr.12: 27368309-27368309 on Build GRCh38
rs943885462 Chr.12: 27368395-27368395 on Build GRCh38
rs764972780 Chr.12: 27368319-27368319 on Build GRCh38
rs753093730 Chr.12: 27368262-27368262 on Build GRCh38
rs776772047 Chr.12: 27368392-27368392 on Build GRCh38
rs981680498 Chr.12: 27368264-27368264 on Build GRCh38
rs759599264 Chr.12: 27368419-27368420 on Build GRCh38
rs764326645 Chr.12: 27368431-27368431 on Build GRCh38
rs772962696 Chr.12: 27368312-27368312 on Build GRCh38
rs781700214 Chr.12: 27368365-27368365 on Build GRCh38
rs770859056 Chr.12: 27368276-27368276 on Build GRCh38
rs771713236 Chr.12: 27368385-27368385 on Build GRCh38
rs781272646 Chr.12: 27368277-27368277 on Build GRCh38
rs761507200 Chr.12: 27368299-27368299 on Build GRCh38
rs747557000 Chr.12: 27368273-27368273 on Build GRCh38
rs757814682 Chr.12: 27368346-27368346 on Build GRCh38
rs756988598 Chr.12: 27368313-27368313 on Build GRCh38
rs751963953 Chr.12: 27368342-27368342 on Build GRCh38
rs778140306 Chr.12: 27368266-27368266 on Build GRCh38
rs780181750 Chr.12: 27368379-27368379 on Build GRCh38
rs745934872 Chr.12: 27368371-27368371 on Build GRCh38
rs746050396 Chr.12: 27368279-27368279 on Build GRCh38
rs759115925 Chr.12: 27368393-27368393 on Build GRCh38
rs775227240 Chr.12: 27368425-27368425 on Build GRCh38
rs769981079 Chr.12: 27368294-27368294 on Build GRCh38
rs377033652 Chr.12: 27368386-27368386 on Build GRCh38
rs531583141 Chr.12: 27368322-27368322 on Build GRCh38
rs764847042 Chr.12: 27368410-27368410 on Build GRCh38
rs111899732 Chr.12: 27368290-27368290 on Build GRCh38

View Full Product Details