Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5206188-5206459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C4
Entrez Gene ID
1109
Gene Name
aldo-keto reductase family 1, member C4
Gene Aliases
3-alpha-HSD, C11, CDR, CHDR, DD-4, DD4, HAKRA
UniGene
Hs.567245
Gene Chromosome Location
Chr.10: 5196835-5218947 on Build GRCh38
Transcripts
NM_001818.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3438575 c.473G>A p.G158E Chr.10: 5206300-5206300 on Build GRCh38
4843140 c.490G>A p.G164R Chr.10: 5206317-5206317 on Build GRCh38
1603511 c.546C>G p.L182L Chr.10: 5206373-5206373 on Build GRCh38
5825443 c.448-2A>G p.? Chr.10: 5206273-5206273 on Build GRCh38
7355128 c.460T>C p.C154R Chr.10: 5206287-5206287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781817169 Chr.10: 5206398-5206398 on Build GRCh38
rs782015173 Chr.10: 5206292-5206292 on Build GRCh38
rs551435912 Chr.10: 5206414-5206414 on Build GRCh38
rs782596820 Chr.10: 5206340-5206340 on Build GRCh38
rs553027648 Chr.10: 5206375-5206375 on Build GRCh38
rs782503549 Chr.10: 5206255-5206255 on Build GRCh38
rs782574528 Chr.10: 5206434-5206434 on Build GRCh38
rs782756855 Chr.10: 5206396-5206396 on Build GRCh38
rs782041453 Chr.10: 5206389-5206389 on Build GRCh38
rs782532788 Chr.10: 5206430-5206430 on Build GRCh38
rs200431502 Chr.10: 5206432-5206432 on Build GRCh38
rs141421379 Chr.10: 5206240-5206240 on Build GRCh38
rs201298644 Chr.10: 5206369-5206369 on Build GRCh38
rs782271542 Chr.10: 5206364-5206364 on Build GRCh38
rs782222141 Chr.10: 5206274-5206274 on Build GRCh38
rs143424974 Chr.10: 5206337-5206337 on Build GRCh38
rs146088459 Chr.10: 5206371-5206371 on Build GRCh38
rs782175017 Chr.10: 5206377-5206377 on Build GRCh38
rs782564763 Chr.10: 5206270-5206270 on Build GRCh38
rs144183289 Chr.10: 5206275-5206275 on Build GRCh38
rs781957701 Chr.10: 5206300-5206300 on Build GRCh38
rs374575386 Chr.10: 5206319-5206319 on Build GRCh38
rs17851824 Chr.10: 5206336-5206336 on Build GRCh38
rs374502554 Chr.10: 5206317-5206317 on Build GRCh38
rs782615208 Chr.10: 5206307-5206308 on Build GRCh38
rs781971093 Chr.10: 5206370-5206370 on Build GRCh38
rs782694538 Chr.10: 5206257-5206257 on Build GRCh38
rs782136434 Chr.10: 5206304-5206304 on Build GRCh38
rs558287231 Chr.10: 5206324-5206324 on Build GRCh38
rs781906133 Chr.10: 5206316-5206316 on Build GRCh38
rs893495516 Chr.10: 5206309-5206309 on Build GRCh38
rs139905834 Chr.10: 5206294-5206294 on Build GRCh38
rs782503904 Chr.10: 5206356-5206356 on Build GRCh38
rs201190050 Chr.10: 5206260-5206260 on Build GRCh38
rs201966709 Chr.10: 5206289-5206289 on Build GRCh38
rs370204291 Chr.10: 5206246-5206246 on Build GRCh38
rs574354508 Chr.10: 5206420-5206420 on Build GRCh38
rs782243403 Chr.10: 5206277-5206277 on Build GRCh38
rs781945929 Chr.10: 5206385-5206385 on Build GRCh38
rs368236673 Chr.10: 5206322-5206322 on Build GRCh38
rs973039053 Chr.10: 5206413-5206413 on Build GRCh38
rs140081411 Chr.10: 5206378-5206378 on Build GRCh38
rs782262285 Chr.10: 5206341-5206341 on Build GRCh38
rs961923110 Chr.10: 5206407-5206407 on Build GRCh38
rs772409192 Chr.10: 5206314-5206314 on Build GRCh38
rs782127048 Chr.10: 5206399-5206399 on Build GRCh38
rs782279849 Chr.10: 5206267-5206267 on Build GRCh38

View Full Product Details