Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203165629-203165882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA1
Entrez Gene ID
134
Gene Name
adenosine A1 receptor
Gene Aliases
RDC7
UniGene
Hs.77867
Gene Chromosome Location
Chr.1: 203127705-203167405 on Build GRCh38
Transcripts
NM_000674.2 NM_001048230.1 XM_005244901.1 XM_005244902.4 XM_017000342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353445 c.934G>A p.A312T Chr.1: 203165853-203165853 on Build GRCh38
5439469 c.799A>G p.S267G Chr.1: 203165718-203165718 on Build GRCh38
6595313 c.744G>C p.L248L Chr.1: 203165663-203165663 on Build GRCh38
2213517 c.737G>C p.S246T Chr.1: 203165656-203165656 on Build GRCh38
5459590 c.837C>A p.G279G Chr.1: 203165756-203165756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200808876 Chr.1: 203165831-203165831 on Build GRCh38
rs772336239 Chr.1: 203165763-203165763 on Build GRCh38
rs752365197 Chr.1: 203165696-203165696 on Build GRCh38
rs766823930 Chr.1: 203165791-203165791 on Build GRCh38
rs781198573 Chr.1: 203165726-203165726 on Build GRCh38
rs200703683 Chr.1: 203165750-203165750 on Build GRCh38
rs1040627926 Chr.1: 203165796-203165796 on Build GRCh38
rs201811129 Chr.1: 203165807-203165807 on Build GRCh38
rs780947937 Chr.1: 203165851-203165851 on Build GRCh38
rs200908360 Chr.1: 203165853-203165853 on Build GRCh38
rs760187303 Chr.1: 203165712-203165712 on Build GRCh38
rs757137013 Chr.1: 203165845-203165845 on Build GRCh38
rs748346254 Chr.1: 203165754-203165754 on Build GRCh38
rs939953705 Chr.1: 203165673-203165673 on Build GRCh38
rs750835106 Chr.1: 203165666-203165666 on Build GRCh38
rs778391614 Chr.1: 203165688-203165688 on Build GRCh38
rs201558854 Chr.1: 203165710-203165710 on Build GRCh38
rs754539453 Chr.1: 203165685-203165685 on Build GRCh38
rs199516537 Chr.1: 203165661-203165661 on Build GRCh38
rs201482062 Chr.1: 203165681-203165681 on Build GRCh38
rs201703869 Chr.1: 203165790-203165790 on Build GRCh38
rs199760746 Chr.1: 203165714-203165714 on Build GRCh38
rs141819948 Chr.1: 203165842-203165842 on Build GRCh38
rs142252167 Chr.1: 203165806-203165806 on Build GRCh38
rs748812754 Chr.1: 203165753-203165753 on Build GRCh38
rs761031787 Chr.1: 203165776-203165776 on Build GRCh38
rs201646414 Chr.1: 203165841-203165841 on Build GRCh38
rs200246307 Chr.1: 203165858-203165858 on Build GRCh38
rs17852405 Chr.1: 203165701-203165701 on Build GRCh38
rs746871603 Chr.1: 203165703-203165703 on Build GRCh38
rs145245940 Chr.1: 203165674-203165674 on Build GRCh38
rs202084025 Chr.1: 203165713-203165713 on Build GRCh38
rs200543088 Chr.1: 203165704-203165704 on Build GRCh38
rs920762648 Chr.1: 203165761-203165761 on Build GRCh38
rs575455405 Chr.1: 203165767-203165767 on Build GRCh38
rs199829860 Chr.1: 203165805-203165805 on Build GRCh38
rs200564867 Chr.1: 203165675-203165675 on Build GRCh38
rs775034018 Chr.1: 203165671-203165671 on Build GRCh38
rs376563906 Chr.1: 203165702-203165702 on Build GRCh38
rs202192831 Chr.1: 203165854-203165854 on Build GRCh38
rs749274582 Chr.1: 203165728-203165728 on Build GRCh38
rs200032682 Chr.1: 203165808-203165808 on Build GRCh38

View Full Product Details