Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 111506237-111506492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STARD4
Entrez Gene ID
134429
Gene Name
StAR related lipid transfer domain containing 4
Gene Aliases
-
UniGene
Hs.732430
Gene Chromosome Location
Chr.5: 111496033-111512597 on Build GRCh38
Transcripts
NM_001308056.1 NM_001308057.1 NM_001308058.1 NM_001308059.1 NM_001308060.1 NM_001308061.1 NM_139164.2 NR_131752.1 NR_131753.1 NR_131754.1 XM_017009041.1 XM_017009042.1 XM_017009043.1 XM_017009044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186808 c.131C>T p.P44L Chr.5: 111506354-111506354 on Build GRCh38
5539779 c.125G>A p.R42K Chr.5: 111506360-111506360 on Build GRCh38
381565 c.112G>A p.V38I Chr.5: 111506373-111506373 on Build GRCh38
3607532 c.139G>A p.E47K Chr.5: 111506346-111506346 on Build GRCh38
2153275 c.150A>C p.G50G Chr.5: 111506335-111506335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765315750 Chr.5: 111506394-111506394 on Build GRCh38
rs904754505 Chr.5: 111506316-111506316 on Build GRCh38
rs749369871 Chr.5: 111506314-111506325 on Build GRCh38
rs1055803258 Chr.5: 111506354-111506354 on Build GRCh38
rs573763173 Chr.5: 111506449-111506449 on Build GRCh38
rs185002486 Chr.5: 111506276-111506276 on Build GRCh38
rs755035924 Chr.5: 111506397-111506398 on Build GRCh38
rs753856103 Chr.5: 111506322-111506325 on Build GRCh38
rs758705998 Chr.5: 111506280-111506280 on Build GRCh38
rs377260389 Chr.5: 111506335-111506335 on Build GRCh38
rs1000872074 Chr.5: 111506396-111506396 on Build GRCh38
rs755614849 Chr.5: 111506411-111506411 on Build GRCh38
rs779775585 Chr.5: 111506413-111506413 on Build GRCh38
rs753138164 Chr.5: 111506395-111506395 on Build GRCh38
rs937095744 Chr.5: 111506275-111506275 on Build GRCh38
rs946177430 Chr.5: 111506299-111506299 on Build GRCh38
rs769446999 Chr.5: 111506315-111506315 on Build GRCh38
rs775507957 Chr.5: 111506323-111506323 on Build GRCh38
rs759564098 Chr.5: 111506385-111506385 on Build GRCh38
rs372000637 Chr.5: 111506415-111506415 on Build GRCh38
rs745327658 Chr.5: 111506311-111506311 on Build GRCh38
rs776869948 Chr.5: 111506379-111506379 on Build GRCh38
rs899859005 Chr.5: 111506427-111506427 on Build GRCh38
rs78019286 Chr.5: 111506451-111506451 on Build GRCh38
rs756831662 Chr.5: 111506399-111506399 on Build GRCh38
rs1045588840 Chr.5: 111506278-111506278 on Build GRCh38
rs777037662 Chr.5: 111506432-111506432 on Build GRCh38
rs764355416 Chr.5: 111506398-111506398 on Build GRCh38
rs746640134 Chr.5: 111506292-111506292 on Build GRCh38
rs774258011 Chr.5: 111506346-111506346 on Build GRCh38
rs759013101 Chr.5: 111506397-111506397 on Build GRCh38
rs372685177 Chr.5: 111506329-111506329 on Build GRCh38
rs756995758 Chr.5: 111506293-111506293 on Build GRCh38
rs779030561 Chr.5: 111506411-111506411 on Build GRCh38
rs773985152 Chr.5: 111506420-111506420 on Build GRCh38
rs1008866080 Chr.5: 111506443-111506443 on Build GRCh38
rs533937023 Chr.5: 111506309-111506309 on Build GRCh38
rs368175619 Chr.5: 111506405-111506405 on Build GRCh38
rs538274131 Chr.5: 111506439-111506439 on Build GRCh38
rs747934896 Chr.5: 111506428-111506428 on Build GRCh38
rs191030185 Chr.5: 111506287-111506287 on Build GRCh38
rs768687390 Chr.5: 111506418-111506418 on Build GRCh38
rs766663499 Chr.5: 111506355-111506355 on Build GRCh38
rs760785187 Chr.5: 111506353-111506353 on Build GRCh38
rs555759618 Chr.5: 111506401-111506401 on Build GRCh38

View Full Product Details