Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18316064-18316297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF133
Entrez Gene ID
7692
Gene Name
zinc finger protein 133
Gene Aliases
ZNF150, pHZ-13, pHZ-66
UniGene
Hs.472221
Gene Chromosome Location
Chr.20: 18288228-18316996 on Build GRCh38
Transcripts
NM_001083330.2 NM_001282995.1 NM_001282996.1 NM_001282997.1 NM_001282998.1 NM_001282999.1 NM_001283000.1 NM_001283001.1 NM_001283002.1 NM_001283003.1 NM_001283004.1 NM_001283005.1 NM_001283006.1 NM_001283007.1 NM_001283008.1 NM_003434.5 XM_005260819.1 XM_005260820.4 XM_011529336.1 XM_011529337.1 XM_011529338.2 XM_011529339.1 XM_011529340.1 XM_017028041.1 XM_017028042.1 XM_017028043.1 XM_017028044.1 XM_017028045.1 XM_017028046.1 XM_017028047.1 XM_017028048.1 XM_017028049.1 XM_017028050.1 XM_017028051.1 XM_017028052.1 XM_017028053.1 XM_017028054.1 XM_017028055.1 XM_017028056.1 XM_017028057.1 XM_017028058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827984 c.1233C>T p.C411C Chr.20: 18316087-18316087 on Build GRCh38
5833932 c.1284_1285insA p.R429fs*19 Chr.20: 18316138-18316139 on Build GRCh38
4581648 c.1365C>T p.H455H Chr.20: 18316219-18316219 on Build GRCh38
7278394 c.1309T>C p.Y437H Chr.20: 18316163-18316163 on Build GRCh38
1713261 c.1321G>T p.V441L Chr.20: 18316175-18316175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373234186 Chr.20: 18316088-18316088 on Build GRCh38
rs775728011 Chr.20: 18316268-18316268 on Build GRCh38
rs764095177 Chr.20: 18316140-18316140 on Build GRCh38
rs770264723 Chr.20: 18316267-18316267 on Build GRCh38
rs766985465 Chr.20: 18316182-18316182 on Build GRCh38
rs762512813 Chr.20: 18316156-18316156 on Build GRCh38
rs115173830 Chr.20: 18316126-18316126 on Build GRCh38
rs200887573 Chr.20: 18316147-18316147 on Build GRCh38
rs748381291 Chr.20: 18316095-18316095 on Build GRCh38
rs541052064 Chr.20: 18316162-18316162 on Build GRCh38
rs746429257 Chr.20: 18316258-18316258 on Build GRCh38
rs200278350 Chr.20: 18316192-18316192 on Build GRCh38
rs144265827 Chr.20: 18316127-18316127 on Build GRCh38
rs775915652 Chr.20: 18316152-18316152 on Build GRCh38
rs2424156 Chr.20: 18316087-18316087 on Build GRCh38
rs773430717 Chr.20: 18316111-18316111 on Build GRCh38
rs540134346 Chr.20: 18316261-18316261 on Build GRCh38
rs777316260 Chr.20: 18316143-18316143 on Build GRCh38
rs146284444 Chr.20: 18316219-18316219 on Build GRCh38
rs755905490 Chr.20: 18316090-18316090 on Build GRCh38
rs757595500 Chr.20: 18316088-18316088 on Build GRCh38
rs148666002 Chr.20: 18316166-18316166 on Build GRCh38
rs772335162 Chr.20: 18316099-18316099 on Build GRCh38
rs757465065 Chr.20: 18316231-18316231 on Build GRCh38
rs143375392 Chr.20: 18316213-18316213 on Build GRCh38
rs781318049 Chr.20: 18316246-18316246 on Build GRCh38
rs747200749 Chr.20: 18316225-18316225 on Build GRCh38
rs34579633 Chr.20: 18316204-18316204 on Build GRCh38
rs779880709 Chr.20: 18316091-18316091 on Build GRCh38
rs376368308 Chr.20: 18316212-18316212 on Build GRCh38
rs758708450 Chr.20: 18316218-18316218 on Build GRCh38
rs781737368 Chr.20: 18316227-18316228 on Build GRCh38
rs760755376 Chr.20: 18316116-18316116 on Build GRCh38
rs559109592 Chr.20: 18316185-18316185 on Build GRCh38
rs763324784 Chr.20: 18316269-18316269 on Build GRCh38
rs751089163 Chr.20: 18316165-18316165 on Build GRCh38
rs146629840 Chr.20: 18316148-18316148 on Build GRCh38
rs746391301 Chr.20: 18316233-18316233 on Build GRCh38

View Full Product Details