Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180950214-180950455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_001159710.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534651.2 XM_011534652.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6170850 c.1339C>T p.P447S Chr.5: 180950380-180950380 on Build GRCh38
1066889 c.1394A>T p.E465V Chr.5: 180950435-180950435 on Build GRCh38
4523625 c.1224G>A p.G408G Chr.5: 180950265-180950265 on Build GRCh38
5683274 c.1223G>C p.G408A Chr.5: 180950264-180950264 on Build GRCh38
737435 c.1388C>A p.T463N Chr.5: 180950429-180950429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992978806 Chr.5: 180950255-180950255 on Build GRCh38
rs775887999 Chr.5: 180950279-180950279 on Build GRCh38
rs1039782700 Chr.5: 180950376-180950376 on Build GRCh38
rs761804981 Chr.5: 180950323-180950323 on Build GRCh38
rs367930588 Chr.5: 180950344-180950344 on Build GRCh38
rs772395850 Chr.5: 180950274-180950274 on Build GRCh38
rs2278689 Chr.5: 180950331-180950331 on Build GRCh38
rs773050693 Chr.5: 180950413-180950413 on Build GRCh38
rs144879081 Chr.5: 180950284-180950284 on Build GRCh38
rs746196868 Chr.5: 180950381-180950381 on Build GRCh38
rs148879045 Chr.5: 180950378-180950378 on Build GRCh38
rs111728368 Chr.5: 180950428-180950428 on Build GRCh38
rs79689901 Chr.5: 180950385-180950385 on Build GRCh38
rs780367262 Chr.5: 180950393-180950393 on Build GRCh38
rs372942569 Chr.5: 180950402-180950402 on Build GRCh38
rs769739300 Chr.5: 180950411-180950411 on Build GRCh38
rs754423182 Chr.5: 180950345-180950345 on Build GRCh38
rs758857333 Chr.5: 180950382-180950382 on Build GRCh38
rs759193395 Chr.5: 180950298-180950300 on Build GRCh38
rs372520525 Chr.5: 180950363-180950363 on Build GRCh38
rs375876659 Chr.5: 180950407-180950407 on Build GRCh38
rs370357739 Chr.5: 180950290-180950290 on Build GRCh38
rs374347303 Chr.5: 180950270-180950270 on Build GRCh38
rs781317825 Chr.5: 180950406-180950406 on Build GRCh38
rs762775062 Chr.5: 180950335-180950335 on Build GRCh38
rs764813890 Chr.5: 180950426-180950427 on Build GRCh38
rs765972831 Chr.5: 180950340-180950340 on Build GRCh38
rs779327087 Chr.5: 180950266-180950266 on Build GRCh38
rs773036543 Chr.5: 180950333-180950333 on Build GRCh38
rs747241268 Chr.5: 180950399-180950399 on Build GRCh38
rs776367234 Chr.5: 180950300-180950300 on Build GRCh38
rs757775137 Chr.5: 180950247-180950247 on Build GRCh38
rs371426432 Chr.5: 180950423-180950423 on Build GRCh38
rs201751538 Chr.5: 180950431-180950431 on Build GRCh38
rs767055822 Chr.5: 180950359-180950359 on Build GRCh38
rs375532895 Chr.5: 180950364-180950364 on Build GRCh38

View Full Product Details