Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22659406-22659679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QB
Entrez Gene ID
713
Gene Name
complement C1q B chain
Gene Aliases
-
UniGene
Hs.8986
Gene Chromosome Location
Chr.1: 22653189-22661536 on Build GRCh38
Transcripts
NM_000491.3 XM_011542059.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667208 c.54C>T p.L18L Chr.1: 22659510-22659510 on Build GRCh38
4643702 c.163A>T p.T55S Chr.1: 22659619-22659619 on Build GRCh38
6385017 c.113C>A p.P38H Chr.1: 22659569-22659569 on Build GRCh38
6124635 c.101_102CC>AA p.P34Q Chr.1: 22659557-22659558 on Build GRCh38
6615369 c.152G>A p.G51D Chr.1: 22659608-22659608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555943495 Chr.1: 22659523-22659523 on Build GRCh38
rs1004187521 Chr.1: 22659589-22659589 on Build GRCh38
rs779270876 Chr.1: 22659487-22659487 on Build GRCh38
rs945618655 Chr.1: 22659588-22659589 on Build GRCh38
rs747307315 Chr.1: 22659593-22659593 on Build GRCh38
rs769378243 Chr.1: 22659508-22659508 on Build GRCh38
rs375854491 Chr.1: 22659430-22659430 on Build GRCh38
rs771991923 Chr.1: 22659625-22659625 on Build GRCh38
rs117009177 Chr.1: 22659444-22659444 on Build GRCh38
rs200249782 Chr.1: 22659556-22659556 on Build GRCh38
rs760529982 Chr.1: 22659639-22659639 on Build GRCh38
rs145215179 Chr.1: 22659579-22659579 on Build GRCh38
rs188344669 Chr.1: 22659470-22659470 on Build GRCh38
rs752493254 Chr.1: 22659525-22659525 on Build GRCh38
rs772289848 Chr.1: 22659499-22659499 on Build GRCh38
rs748847255 Chr.1: 22659603-22659603 on Build GRCh38
rs184783763 Chr.1: 22659641-22659641 on Build GRCh38
rs746291318 Chr.1: 22659498-22659498 on Build GRCh38
rs751439188 Chr.1: 22659428-22659428 on Build GRCh38
rs147623518 Chr.1: 22659618-22659618 on Build GRCh38
rs778026557 Chr.1: 22659445-22659445 on Build GRCh38
rs753914766 Chr.1: 22659453-22659453 on Build GRCh38
rs775434206 Chr.1: 22659630-22659630 on Build GRCh38
rs375361665 Chr.1: 22659514-22659514 on Build GRCh38
rs762052370 Chr.1: 22659646-22659646 on Build GRCh38
rs780407647 Chr.1: 22659588-22659588 on Build GRCh38
rs77379868 Chr.1: 22659522-22659522 on Build GRCh38
rs770317321 Chr.1: 22659604-22659604 on Build GRCh38
rs1057480674 Chr.1: 22659515-22659515 on Build GRCh38
rs757258422 Chr.1: 22659561-22659561 on Build GRCh38
rs747818615 Chr.1: 22659506-22659506 on Build GRCh38
rs764456393 Chr.1: 22659640-22659640 on Build GRCh38
rs773875225 Chr.1: 22659612-22659612 on Build GRCh38
rs1024240579 Chr.1: 22659435-22659435 on Build GRCh38
rs951341286 Chr.1: 22659469-22659469 on Build GRCh38
rs765281728 Chr.1: 22659617-22659617 on Build GRCh38
rs777391100 Chr.1: 22659601-22659601 on Build GRCh38
rs769504566 Chr.1: 22659596-22659596 on Build GRCh38
rs750922119 Chr.1: 22659566-22659566 on Build GRCh38
rs909697564 Chr.1: 22659502-22659502 on Build GRCh38
rs764348361 Chr.1: 22659553-22659553 on Build GRCh38
rs142278562 Chr.1: 22659564-22659564 on Build GRCh38
rs866902627 Chr.1: 22659482-22659482 on Build GRCh38
rs780425216 Chr.1: 22659501-22659501 on Build GRCh38
rs748217347 Chr.1: 22659552-22659552 on Build GRCh38
rs534264704 Chr.1: 22659443-22659443 on Build GRCh38
rs760994723 Chr.1: 22659551-22659551 on Build GRCh38

View Full Product Details