Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 19473175-19473442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP2
Entrez Gene ID
121536
Gene Name
AE binding protein 2
Gene Aliases
-
UniGene
Hs.126497
Gene Chromosome Location
Chr.12: 19404101-19522239 on Build GRCh38
Transcripts
NM_001114176.1 NM_001267043.1 NM_153207.4 XM_006719031.3 XM_017018806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5041704 c.900A>T p.K300N Chr.12: 19473268-19473268 on Build GRCh38
4660171 c.969T>C p.S323S Chr.12: 19473337-19473337 on Build GRCh38
7354660 c.253G>T p.G85C Chr.12: 19473269-19473269 on Build GRCh38
7246164 c.320G>A p.S107N Chr.12: 19473336-19473336 on Build GRCh38
1238141 c.958C>A p.L320M Chr.12: 19473326-19473326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771914756 Chr.12: 19473348-19473348 on Build GRCh38
rs761293040 Chr.12: 19473376-19473377 on Build GRCh38
rs747185168 Chr.12: 19473262-19473262 on Build GRCh38
rs12818887 Chr.12: 19473396-19473396 on Build GRCh38
rs11044583 Chr.12: 19473388-19473388 on Build GRCh38
rs772744354 Chr.12: 19473366-19473366 on Build GRCh38
rs369601142 Chr.12: 19473388-19473388 on Build GRCh38
rs770366287 Chr.12: 19473371-19473371 on Build GRCh38
rs57817761 Chr.12: 19473390-19473392 on Build GRCh38
rs746103128 Chr.12: 19473382-19473396 on Build GRCh38
rs781682898 Chr.12: 19473292-19473292 on Build GRCh38
rs780606365 Chr.12: 19473337-19473337 on Build GRCh38
rs765045716 Chr.12: 19473205-19473205 on Build GRCh38
rs71530937 Chr.12: 19473400-19473400 on Build GRCh38
rs545170365 Chr.12: 19473334-19473334 on Build GRCh38
rs565267420 Chr.12: 19473369-19473369 on Build GRCh38
rs56979677 Chr.12: 19473398-19473400 on Build GRCh38
rs777819791 Chr.12: 19473243-19473243 on Build GRCh38
rs546038420 Chr.12: 19473380-19473381 on Build GRCh38
rs10631217 Chr.12: 19473379-19473380 on Build GRCh38
rs542134245 Chr.12: 19473382-19473384 on Build GRCh38
rs10631218 Chr.12: 19473380-19473380 on Build GRCh38
rs779179164 Chr.12: 19473377-19473384 on Build GRCh38
rs779195002 Chr.12: 19473378-19473378 on Build GRCh38
rs750173802 Chr.12: 19473392-19473393 on Build GRCh38
rs182141135 Chr.12: 19473404-19473404 on Build GRCh38
rs758671245 Chr.12: 19473234-19473234 on Build GRCh38
rs201557773 Chr.12: 19473378-19473379 on Build GRCh38
rs751212235 Chr.12: 19473377-19473380 on Build GRCh38
rs777864449 Chr.12: 19473374-19473375 on Build GRCh38
rs747788509 Chr.12: 19473345-19473345 on Build GRCh38
rs752549686 Chr.12: 19473385-19473385 on Build GRCh38
rs775739294 Chr.12: 19473381-19473382 on Build GRCh38
rs772142531 Chr.12: 19473398-19473399 on Build GRCh38
rs746384559 Chr.12: 19473316-19473316 on Build GRCh38
rs57423687 Chr.12: 19473384-19473384 on Build GRCh38
rs760945177 Chr.12: 19473381-19473384 on Build GRCh38
rs370128326 Chr.12: 19473222-19473224 on Build GRCh38
rs375681057 Chr.12: 19473208-19473208 on Build GRCh38
rs56666201 Chr.12: 19473271-19473271 on Build GRCh38
rs1019409763 Chr.12: 19473219-19473219 on Build GRCh38
rs199934534 Chr.12: 19473375-19473376 on Build GRCh38
rs778796461 Chr.12: 19473382-19473392 on Build GRCh38
rs866412180 Chr.12: 19473408-19473408 on Build GRCh38
rs757358045 Chr.12: 19473382-19473388 on Build GRCh38
rs57086852 Chr.12: 19473396-19473396 on Build GRCh38
rs12370241 Chr.12: 19473392-19473392 on Build GRCh38

View Full Product Details