Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2692250-2692485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUH
Entrez Gene ID
23277
Gene Name
clustered mitochondria homolog
Gene Aliases
CLU1, KIAA0664
UniGene
Hs.22616
Gene Chromosome Location
Chr.17: 2689386-2711633 on Build GRCh38
Transcripts
NM_015229.3 XM_011523768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
302475 c.3331G>A p.G1111R Chr.17: 2692473-2692473 on Build GRCh38
6867570 c.3347G>T p.G1116V Chr.17: 2692457-2692457 on Build GRCh38
4695336 c.3371T>C p.L1124P Chr.17: 2692433-2692433 on Build GRCh38
4064826 c.3374G>A p.R1125H Chr.17: 2692430-2692430 on Build GRCh38
3969903 c.3347G>A p.G1116E Chr.17: 2692457-2692457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537910876 Chr.17: 2692351-2692351 on Build GRCh38
rs754189291 Chr.17: 2692399-2692399 on Build GRCh38
rs772945474 Chr.17: 2692350-2692350 on Build GRCh38
rs1042086810 Chr.17: 2692435-2692435 on Build GRCh38
rs769550827 Chr.17: 2692347-2692347 on Build GRCh38
rs867497496 Chr.17: 2692362-2692362 on Build GRCh38
rs759649141 Chr.17: 2692389-2692389 on Build GRCh38
rs539095875 Chr.17: 2692278-2692278 on Build GRCh38
rs961961374 Chr.17: 2692413-2692413 on Build GRCh38
rs924870904 Chr.17: 2692299-2692299 on Build GRCh38
rs61750787 Chr.17: 2692409-2692409 on Build GRCh38
rs1024845010 Chr.17: 2692294-2692295 on Build GRCh38
rs781037770 Chr.17: 2692411-2692411 on Build GRCh38
rs768530755 Chr.17: 2692337-2692337 on Build GRCh38
rs764583337 Chr.17: 2692396-2692396 on Build GRCh38
rs771077108 Chr.17: 2692373-2692373 on Build GRCh38
rs760299805 Chr.17: 2692307-2692307 on Build GRCh38
rs1054668792 Chr.17: 2692329-2692329 on Build GRCh38
rs763197154 Chr.17: 2692358-2692358 on Build GRCh38
rs374574637 Chr.17: 2692410-2692410 on Build GRCh38
rs774431181 Chr.17: 2692387-2692387 on Build GRCh38
rs760995859 Chr.17: 2692394-2692394 on Build GRCh38
rs747930199 Chr.17: 2692343-2692343 on Build GRCh38
rs35346201 Chr.17: 2692321-2692321 on Build GRCh38
rs777551831 Chr.17: 2692442-2692442 on Build GRCh38
rs368522006 Chr.17: 2692465-2692465 on Build GRCh38
rs932284495 Chr.17: 2692391-2692391 on Build GRCh38
rs372503652 Chr.17: 2692405-2692405 on Build GRCh38
rs558760283 Chr.17: 2692288-2692288 on Build GRCh38
rs748143342 Chr.17: 2692423-2692423 on Build GRCh38
rs936219138 Chr.17: 2692317-2692317 on Build GRCh38
rs377102362 Chr.17: 2692408-2692408 on Build GRCh38
rs781182285 Chr.17: 2692339-2692339 on Build GRCh38
rs769683584 Chr.17: 2692429-2692429 on Build GRCh38
rs768471157 Chr.17: 2692346-2692346 on Build GRCh38
rs746469206 Chr.17: 2692313-2692313 on Build GRCh38
rs994713642 Chr.17: 2692417-2692417 on Build GRCh38
rs1012081415 Chr.17: 2692285-2692285 on Build GRCh38
rs767977011 Chr.17: 2692390-2692390 on Build GRCh38

View Full Product Details