Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 66123779-66124022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LLPH
Entrez Gene ID
84298
Gene Name
LLP homolog, long-term synaptic facilitation
Gene Aliases
C12orf31, hLLP
UniGene
Hs.504820
Gene Chromosome Location
Chr.12: 66123069-66130753 on Build GRCh38
Transcripts
NM_032338.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
459395 c.346A>C p.S116R Chr.12: 66123884-66123884 on Build GRCh38
1991774 c.346delA p.S116fs*6 Chr.12: 66123884-66123884 on Build GRCh38
468784 c.346A>G p.S116G Chr.12: 66123884-66123884 on Build GRCh38
1991775 c.335G>T p.R112I Chr.12: 66123895-66123895 on Build GRCh38
6701641 c.326G>A p.R109Q Chr.12: 66123904-66123904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775187482 Chr.12: 66123906-66123906 on Build GRCh38
rs758166357 Chr.12: 66123982-66123982 on Build GRCh38
rs761418149 Chr.12: 66123953-66123953 on Build GRCh38
rs750421842 Chr.12: 66123966-66123966 on Build GRCh38
rs770343123 Chr.12: 66123898-66123898 on Build GRCh38
rs906913142 Chr.12: 66123915-66123915 on Build GRCh38
rs747729087 Chr.12: 66123892-66123892 on Build GRCh38
rs562308472 Chr.12: 66123909-66123909 on Build GRCh38
rs757332900 Chr.12: 66123844-66123844 on Build GRCh38
rs1054375611 Chr.12: 66123815-66123815 on Build GRCh38
rs532313455 Chr.12: 66123843-66123853 on Build GRCh38
rs773722277 Chr.12: 66123901-66123901 on Build GRCh38
rs748038311 Chr.12: 66123829-66123830 on Build GRCh38
rs373853440 Chr.12: 66123833-66123834 on Build GRCh38
rs548981790 Chr.12: 66123860-66123860 on Build GRCh38
rs945887029 Chr.12: 66123878-66123878 on Build GRCh38
rs201431090 Chr.12: 66123847-66123847 on Build GRCh38
rs755059967 Chr.12: 66123870-66123870 on Build GRCh38
rs773003450 Chr.12: 66123888-66123889 on Build GRCh38
rs966513113 Chr.12: 66123824-66123824 on Build GRCh38
rs751777131 Chr.12: 66123995-66123995 on Build GRCh38
rs771222748 Chr.12: 66123905-66123905 on Build GRCh38
rs1001105569 Chr.12: 66123894-66123894 on Build GRCh38
rs763653722 Chr.12: 66123938-66123938 on Build GRCh38
rs147676166 Chr.12: 66123947-66123947 on Build GRCh38
rs762847338 Chr.12: 66123920-66123920 on Build GRCh38
rs753779731 Chr.12: 66123823-66123823 on Build GRCh38
rs758277426 Chr.12: 66123853-66123853 on Build GRCh38
rs1033902585 Chr.12: 66123929-66123929 on Build GRCh38
rs749613152 Chr.12: 66123904-66123904 on Build GRCh38
rs368405358 Chr.12: 66123890-66123890 on Build GRCh38
rs766139385 Chr.12: 66123994-66123994 on Build GRCh38
rs763562496 Chr.12: 66123810-66123810 on Build GRCh38
rs1034135315 Chr.12: 66123949-66123949 on Build GRCh38
rs765196322 Chr.12: 66123962-66123962 on Build GRCh38

View Full Product Details