Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24181528-24181783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
UniGene
Hs.411865
Gene Chromosome Location
Chr.14: 24180219-24188918 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
327450 c.2994G>A p.W998* Chr.14: 24181564-24181564 on Build GRCh38
4845867 c.2936C>T p.P979L Chr.14: 24181715-24181715 on Build GRCh38
1159576 c.2873G>C p.R958P Chr.14: 24181778-24181778 on Build GRCh38
3983205 c.2980G>A p.D994N Chr.14: 24181578-24181578 on Build GRCh38
4643832 c.2893C>G p.L965V Chr.14: 24181758-24181758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769441387 Chr.14: 24181744-24181744 on Build GRCh38
rs770526078 Chr.14: 24181759-24181759 on Build GRCh38
rs748753318 Chr.14: 24181719-24181719 on Build GRCh38
rs980374013 Chr.14: 24181602-24181602 on Build GRCh38
rs774983003 Chr.14: 24181663-24181663 on Build GRCh38
rs138213962 Chr.14: 24181704-24181704 on Build GRCh38
rs747836677 Chr.14: 24181619-24181619 on Build GRCh38
rs774821596 Chr.14: 24181698-24181701 on Build GRCh38
rs767968161 Chr.14: 24181670-24181670 on Build GRCh38
rs746913932 Chr.14: 24181596-24181596 on Build GRCh38
rs937705958 Chr.14: 24181626-24181626 on Build GRCh38
rs547693598 Chr.14: 24181717-24181717 on Build GRCh38
rs958410864 Chr.14: 24181551-24181551 on Build GRCh38
rs769396452 Chr.14: 24181621-24181621 on Build GRCh38
rs753015395 Chr.14: 24181555-24181555 on Build GRCh38
rs199744643 Chr.14: 24181695-24181695 on Build GRCh38
rs756375329 Chr.14: 24181556-24181556 on Build GRCh38
rs760122991 Chr.14: 24181762-24181762 on Build GRCh38
rs758419289 Chr.14: 24181574-24181574 on Build GRCh38
rs768550676 Chr.14: 24181605-24181605 on Build GRCh38
rs762383520 Chr.14: 24181630-24181630 on Build GRCh38
rs374679303 Chr.14: 24181589-24181589 on Build GRCh38
rs200789473 Chr.14: 24181710-24181710 on Build GRCh38
rs147005860 Chr.14: 24181624-24181624 on Build GRCh38
rs765870338 Chr.14: 24181631-24181631 on Build GRCh38
rs752967543 Chr.14: 24181674-24181674 on Build GRCh38
rs748926988 Chr.14: 24181753-24181753 on Build GRCh38
rs781070992 Chr.14: 24181731-24181731 on Build GRCh38
rs764358305 Chr.14: 24181700-24181700 on Build GRCh38
rs754029210 Chr.14: 24181666-24181666 on Build GRCh38
rs777470214 Chr.14: 24181751-24181751 on Build GRCh38
rs372882135 Chr.14: 24181573-24181573 on Build GRCh38
rs778034805 Chr.14: 24181558-24181558 on Build GRCh38
rs747889812 Chr.14: 24181734-24181734 on Build GRCh38
rs1038144935 Chr.14: 24181752-24181752 on Build GRCh38
rs770452054 Chr.14: 24181565-24181566 on Build GRCh38
rs369101802 Chr.14: 24181550-24181550 on Build GRCh38
rs778843210 Chr.14: 24181715-24181715 on Build GRCh38
rs776266326 Chr.14: 24181607-24181607 on Build GRCh38
rs59484787 Chr.14: 24181761-24181761 on Build GRCh38
rs759872782 Chr.14: 24181752-24181752 on Build GRCh38

View Full Product Details