Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153862730-153863003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L1CAM
Entrez Gene ID
3897
Gene Name
L1 cell adhesion molecule
Gene Aliases
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1
UniGene
Hs.522818
Gene Chromosome Location
Chr.X: 153861514-153886174 on Build GRCh38
Transcripts
NM_000425.4 NM_001143963.2 NM_001278116.1 NM_024003.3

SNP Detail

SNP ID Chromosome Location
rs200235262 Chr.X: 153862828-153862828 on Build GRCh38
rs782678040 Chr.X: 153862864-153862864 on Build GRCh38
rs201732197 Chr.X: 153862909-153862909 on Build GRCh38
rs189562799 Chr.X: 153862970-153862970 on Build GRCh38
rs782119181 Chr.X: 153862934-153862934 on Build GRCh38
rs373984208 Chr.X: 153862836-153862836 on Build GRCh38
rs782702177 Chr.X: 153862842-153862842 on Build GRCh38
rs199810233 Chr.X: 153862837-153862837 on Build GRCh38
rs782110886 Chr.X: 153862822-153862822 on Build GRCh38
rs782341083 Chr.X: 153862795-153862795 on Build GRCh38
rs376315514 Chr.X: 153862858-153862858 on Build GRCh38
rs148687917 Chr.X: 153862865-153862865 on Build GRCh38
rs200916591 Chr.X: 153862888-153862888 on Build GRCh38
rs782225211 Chr.X: 153862905-153862905 on Build GRCh38
rs782028847 Chr.X: 153862945-153862945 on Build GRCh38
rs781858962 Chr.X: 153862855-153862855 on Build GRCh38
rs199870210 Chr.X: 153862923-153862923 on Build GRCh38
rs370597765 Chr.X: 153862910-153862910 on Build GRCh38
rs781923954 Chr.X: 153862918-153862918 on Build GRCh38
rs782716610 Chr.X: 153862975-153862975 on Build GRCh38
rs782563171 Chr.X: 153862780-153862780 on Build GRCh38
rs137852522 Chr.X: 153862856-153862856 on Build GRCh38
rs782086274 Chr.X: 153862849-153862849 on Build GRCh38
rs782388610 Chr.X: 153862777-153862777 on Build GRCh38
rs782774486 Chr.X: 153862941-153862941 on Build GRCh38
rs781784745 Chr.X: 153862868-153862868 on Build GRCh38
rs782366295 Chr.X: 153862907-153862907 on Build GRCh38
rs201625971 Chr.X: 153862762-153862762 on Build GRCh38
rs781814421 Chr.X: 153862848-153862848 on Build GRCh38
rs782423059 Chr.X: 153862917-153862917 on Build GRCh38
rs782351247 Chr.X: 153862924-153862924 on Build GRCh38
rs782167954 Chr.X: 153862838-153862838 on Build GRCh38
rs782405837 Chr.X: 153862898-153862898 on Build GRCh38
rs200931972 Chr.X: 153862933-153862933 on Build GRCh38
rs782020341 Chr.X: 153862831-153862831 on Build GRCh38
rs782191514 Chr.X: 153862787-153862787 on Build GRCh38
rs199975913 Chr.X: 153862916-153862916 on Build GRCh38
rs146526273 Chr.X: 153862765-153862765 on Build GRCh38
rs781964622 Chr.X: 153862809-153862809 on Build GRCh38

View Full Product Details