Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35131611-35131842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDEM2
Entrez Gene ID
55741
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Gene Aliases
C20orf31, C20orf49, bA4204.1
UniGene
Hs.720177
Gene Chromosome Location
Chr.20: 35115357-35147358 on Build GRCh38
Transcripts
NM_001145025.1 NM_018217.2 NR_026728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204913 c.797A>G p.K266R Chr.20: 35131689-35131689 on Build GRCh38
6651585 c.599T>C p.V200A Chr.20: 35131764-35131764 on Build GRCh38
3545643 c.833C>T p.A278V Chr.20: 35131653-35131653 on Build GRCh38
7182133 c.799G>A p.G267R Chr.20: 35131687-35131687 on Build GRCh38
1242219 c.805A>G p.I269V Chr.20: 35131681-35131681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760148994 Chr.20: 35131738-35131738 on Build GRCh38
rs971687140 Chr.20: 35131723-35131723 on Build GRCh38
rs927097611 Chr.20: 35131735-35131735 on Build GRCh38
rs780240357 Chr.20: 35131785-35131785 on Build GRCh38
rs751814252 Chr.20: 35131772-35131772 on Build GRCh38
rs146389165 Chr.20: 35131676-35131676 on Build GRCh38
rs535462035 Chr.20: 35131695-35131695 on Build GRCh38
rs754810516 Chr.20: 35131797-35131797 on Build GRCh38
rs1050588838 Chr.20: 35131727-35131727 on Build GRCh38
rs767630914 Chr.20: 35131776-35131776 on Build GRCh38
rs139566342 Chr.20: 35131736-35131736 on Build GRCh38
rs112207352 Chr.20: 35131757-35131757 on Build GRCh38
rs201265564 Chr.20: 35131734-35131734 on Build GRCh38
rs562242351 Chr.20: 35131814-35131814 on Build GRCh38
rs745930445 Chr.20: 35131656-35131656 on Build GRCh38
rs751838689 Chr.20: 35131636-35131636 on Build GRCh38
rs944977009 Chr.20: 35131753-35131753 on Build GRCh38
rs994971704 Chr.20: 35131655-35131655 on Build GRCh38
rs1042360259 Chr.20: 35131800-35131800 on Build GRCh38
rs781346888 Chr.20: 35131642-35131642 on Build GRCh38
rs776003033 Chr.20: 35131770-35131770 on Build GRCh38
rs143005174 Chr.20: 35131740-35131740 on Build GRCh38
rs762154079 Chr.20: 35131774-35131774 on Build GRCh38
rs377575587 Chr.20: 35131639-35131639 on Build GRCh38
rs778823343 Chr.20: 35131803-35131803 on Build GRCh38
rs753830818 Chr.20: 35131788-35131788 on Build GRCh38
rs771607122 Chr.20: 35131726-35131726 on Build GRCh38
rs576050427 Chr.20: 35131718-35131718 on Build GRCh38
rs768526242 Chr.20: 35131697-35131697 on Build GRCh38
rs760031914 Chr.20: 35131813-35131813 on Build GRCh38
rs200083675 Chr.20: 35131780-35131780 on Build GRCh38
rs749188020 Chr.20: 35131679-35131679 on Build GRCh38
rs199686056 Chr.20: 35131717-35131717 on Build GRCh38
rs763413780 Chr.20: 35131771-35131771 on Build GRCh38
rs377175960 Chr.20: 35131657-35131657 on Build GRCh38
rs147052475 Chr.20: 35131805-35131805 on Build GRCh38
rs146048652 Chr.20: 35131781-35131781 on Build GRCh38
rs573496754 Chr.20: 35131804-35131804 on Build GRCh38
rs983114403 Chr.20: 35131700-35131700 on Build GRCh38

View Full Product Details