Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9859843-9860098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLP2R
Entrez Gene ID
9340
Gene Name
glucagon like peptide 2 receptor
Gene Aliases
-
UniGene
Hs.248202
Gene Chromosome Location
Chr.17: 9825508-9894163 on Build GRCh38
Transcripts
NM_004246.2 XM_005256861.3 XM_011524077.2 XM_017025339.1 XM_017025340.1 XM_017025341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370990 c.892C>T p.R298W Chr.17: 9860068-9860068 on Build GRCh38
986470 c.895C>A p.L299M Chr.17: 9860071-9860071 on Build GRCh38
5671121 c.858C>T p.H286H Chr.17: 9860034-9860034 on Build GRCh38
708473 c.782G>C p.R261P Chr.17: 9859958-9859958 on Build GRCh38
986469 c.872C>T p.P291L Chr.17: 9860048-9860048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776764466 Chr.17: 9859907-9859907 on Build GRCh38
rs140277329 Chr.17: 9860023-9860023 on Build GRCh38
rs750850540 Chr.17: 9859932-9859932 on Build GRCh38
rs754665708 Chr.17: 9859958-9859958 on Build GRCh38
rs114273181 Chr.17: 9860069-9860069 on Build GRCh38
rs978533852 Chr.17: 9859895-9859895 on Build GRCh38
rs780635088 Chr.17: 9859989-9859989 on Build GRCh38
rs146420890 Chr.17: 9860037-9860037 on Build GRCh38
rs1047334603 Chr.17: 9859866-9859866 on Build GRCh38
rs762263243 Chr.17: 9859926-9859926 on Build GRCh38
rs6503255 Chr.17: 9859882-9859882 on Build GRCh38
rs199908559 Chr.17: 9860064-9860064 on Build GRCh38
rs537869166 Chr.17: 9859887-9859887 on Build GRCh38
rs969805754 Chr.17: 9859888-9859888 on Build GRCh38
rs148657181 Chr.17: 9859952-9859952 on Build GRCh38
rs151178828 Chr.17: 9859990-9859990 on Build GRCh38
rs747295129 Chr.17: 9860031-9860031 on Build GRCh38
rs145002314 Chr.17: 9859950-9859950 on Build GRCh38
rs770672135 Chr.17: 9859903-9859903 on Build GRCh38
rs115632347 Chr.17: 9859904-9859904 on Build GRCh38
rs765756568 Chr.17: 9859927-9859927 on Build GRCh38
rs567281503 Chr.17: 9859883-9859883 on Build GRCh38
rs999158778 Chr.17: 9859947-9859947 on Build GRCh38
rs774116566 Chr.17: 9860025-9860025 on Build GRCh38
rs749154873 Chr.17: 9860011-9860011 on Build GRCh38
rs766910205 Chr.17: 9860068-9860068 on Build GRCh38
rs142122440 Chr.17: 9859957-9859957 on Build GRCh38
rs145296340 Chr.17: 9860036-9860036 on Build GRCh38
rs117589128 Chr.17: 9859881-9859881 on Build GRCh38
rs759708925 Chr.17: 9860074-9860074 on Build GRCh38
rs888492974 Chr.17: 9860074-9860074 on Build GRCh38
rs187679366 Chr.17: 9860077-9860077 on Build GRCh38
rs773777004 Chr.17: 9860040-9860040 on Build GRCh38
rs765195268 Chr.17: 9860039-9860039 on Build GRCh38
rs930199270 Chr.17: 9859865-9859865 on Build GRCh38
rs763497998 Chr.17: 9860054-9860054 on Build GRCh38
rs566764219 Chr.17: 9859867-9859867 on Build GRCh38
rs772929795 Chr.17: 9859896-9859896 on Build GRCh38
rs777796266 Chr.17: 9860001-9860001 on Build GRCh38
rs902883437 Chr.17: 9859924-9859924 on Build GRCh38
rs72822175 Chr.17: 9859899-9859899 on Build GRCh38
rs755529488 Chr.17: 9859991-9859991 on Build GRCh38
rs758867906 Chr.17: 9859942-9859942 on Build GRCh38

View Full Product Details