Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37461899-37462172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR1
Entrez Gene ID
23070
Gene Name
cap methyltransferase 1
Gene Aliases
FTSJD2, KIAA0082, MTr1, hMTr1
UniGene
Hs.520102
Gene Chromosome Location
Chr.6: 37433131-37482844 on Build GRCh38
Transcripts
NM_015050.2 XM_005248955.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1078676 c.1276C>T p.R426* Chr.6: 37462053-37462053 on Build GRCh38
4463337 c.1287C>T p.L429L Chr.6: 37462064-37462064 on Build GRCh38
5531500 c.1227C>T p.F409F Chr.6: 37462004-37462004 on Build GRCh38
6012264 c.1242G>C p.V414V Chr.6: 37462019-37462019 on Build GRCh38
3941724 c.1251C>T p.V417V Chr.6: 37462028-37462028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750060888 Chr.6: 37461948-37461948 on Build GRCh38
rs752951174 Chr.6: 37462084-37462084 on Build GRCh38
rs758786979 Chr.6: 37461997-37461997 on Build GRCh38
rs765490947 Chr.6: 37461980-37461980 on Build GRCh38
rs753139757 Chr.6: 37461995-37461995 on Build GRCh38
rs777553421 Chr.6: 37461961-37461961 on Build GRCh38
rs372462143 Chr.6: 37462109-37462109 on Build GRCh38
rs556310068 Chr.6: 37461959-37461959 on Build GRCh38
rs576689024 Chr.6: 37462039-37462039 on Build GRCh38
rs749539327 Chr.6: 37462141-37462141 on Build GRCh38
rs756231041 Chr.6: 37462129-37462129 on Build GRCh38
rs778441597 Chr.6: 37461999-37461999 on Build GRCh38
rs538997957 Chr.6: 37462091-37462091 on Build GRCh38
rs768410267 Chr.6: 37462038-37462038 on Build GRCh38
rs750537797 Chr.6: 37462121-37462121 on Build GRCh38
rs1031209046 Chr.6: 37462143-37462143 on Build GRCh38
rs148875465 Chr.6: 37462076-37462076 on Build GRCh38
rs757408510 Chr.6: 37462006-37462006 on Build GRCh38
rs768422988 Chr.6: 37462148-37462148 on Build GRCh38
rs773792149 Chr.6: 37461934-37461934 on Build GRCh38
rs764112328 Chr.6: 37461925-37461925 on Build GRCh38
rs781482887 Chr.6: 37462022-37462022 on Build GRCh38
rs779662605 Chr.6: 37462028-37462028 on Build GRCh38
rs749121526 Chr.6: 37462032-37462032 on Build GRCh38
rs766080515 Chr.6: 37462083-37462083 on Build GRCh38
rs751624584 Chr.6: 37462005-37462005 on Build GRCh38
rs770084774 Chr.6: 37462026-37462026 on Build GRCh38
rs761316047 Chr.6: 37461944-37461944 on Build GRCh38
rs772745947 Chr.6: 37462054-37462054 on Build GRCh38
rs771545316 Chr.6: 37462053-37462053 on Build GRCh38
rs757889361 Chr.6: 37462113-37462113 on Build GRCh38
rs184268416 Chr.6: 37462120-37462120 on Build GRCh38
rs780327343 Chr.6: 37462132-37462132 on Build GRCh38

View Full Product Details