Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19382159-19382424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK7
Entrez Gene ID
5598
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
BMK1, ERK4, ERK5, PRKM7
UniGene
Hs.150136
Gene Chromosome Location
Chr.17: 19377721-19383544 on Build GRCh38
Transcripts
NM_002749.3 NM_139032.2 NM_139033.2 NM_139034.2 XM_006721557.3 XM_006721558.3 XM_006721559.3 XM_011523957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6002979 c.2052G>T p.L684F Chr.17: 19382355-19382355 on Build GRCh38
4883765 c.1903C>A p.P635T Chr.17: 19382206-19382206 on Build GRCh38
5929740 c.2093G>A p.G698E Chr.17: 19382396-19382396 on Build GRCh38
3515033 c.1920C>T p.P640P Chr.17: 19382223-19382223 on Build GRCh38
6707873 c.1977C>T p.T659T Chr.17: 19382280-19382280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753795731 Chr.17: 19382319-19382319 on Build GRCh38
rs374391055 Chr.17: 19382290-19382290 on Build GRCh38
rs768990208 Chr.17: 19382184-19382184 on Build GRCh38
rs184241833 Chr.17: 19382388-19382388 on Build GRCh38
rs769168649 Chr.17: 19382205-19382205 on Build GRCh38
rs201426673 Chr.17: 19382308-19382308 on Build GRCh38
rs772589753 Chr.17: 19382259-19382259 on Build GRCh38
rs200536560 Chr.17: 19382285-19382285 on Build GRCh38
rs766353128 Chr.17: 19382315-19382315 on Build GRCh38
rs771523538 Chr.17: 19382262-19382262 on Build GRCh38
rs367852843 Chr.17: 19382390-19382390 on Build GRCh38
rs144078337 Chr.17: 19382260-19382260 on Build GRCh38
rs768130772 Chr.17: 19382378-19382378 on Build GRCh38
rs199558212 Chr.17: 19382299-19382299 on Build GRCh38
rs377122950 Chr.17: 19382180-19382180 on Build GRCh38
rs781736991 Chr.17: 19382364-19382364 on Build GRCh38
rs750284588 Chr.17: 19382304-19382304 on Build GRCh38
rs762496317 Chr.17: 19382210-19382210 on Build GRCh38
rs866948388 Chr.17: 19382197-19382197 on Build GRCh38
rs763816343 Chr.17: 19382212-19382212 on Build GRCh38
rs773889944 Chr.17: 19382379-19382379 on Build GRCh38
rs772642554 Chr.17: 19382194-19382194 on Build GRCh38
rs199788725 Chr.17: 19382323-19382323 on Build GRCh38
rs1007543565 Chr.17: 19382342-19382342 on Build GRCh38
rs972699681 Chr.17: 19382389-19382389 on Build GRCh38
rs1001125878 Chr.17: 19382331-19382331 on Build GRCh38
rs748539235 Chr.17: 19382191-19382191 on Build GRCh38
rs759070915 Chr.17: 19382202-19382202 on Build GRCh38
rs1017349815 Chr.17: 19382358-19382358 on Build GRCh38
rs371008501 Chr.17: 19382218-19382218 on Build GRCh38
rs776655771 Chr.17: 19382397-19382397 on Build GRCh38
rs758738832 Chr.17: 19382340-19382340 on Build GRCh38
rs147306640 Chr.17: 19382346-19382346 on Build GRCh38
rs774014133 Chr.17: 19382294-19382294 on Build GRCh38
rs868484799 Chr.17: 19382314-19382314 on Build GRCh38
rs754008504 Chr.17: 19382237-19382237 on Build GRCh38
rs372317367 Chr.17: 19382336-19382336 on Build GRCh38
rs554667705 Chr.17: 19382234-19382234 on Build GRCh38
rs138826180 Chr.17: 19382392-19382392 on Build GRCh38
rs747794000 Chr.17: 19382381-19382381 on Build GRCh38
rs761499978 Chr.17: 19382296-19382296 on Build GRCh38
rs767392187 Chr.17: 19382225-19382225 on Build GRCh38
rs749056959 Chr.17: 19382261-19382261 on Build GRCh38
rs958886505 Chr.17: 19382338-19382338 on Build GRCh38
rs750337390 Chr.17: 19382232-19382232 on Build GRCh38
rs1042293921 Chr.17: 19382289-19382289 on Build GRCh38
rs755099667 Chr.17: 19382240-19382240 on Build GRCh38
rs771271773 Chr.17: 19382361-19382361 on Build GRCh38
rs760525418 Chr.17: 19382391-19382391 on Build GRCh38
rs755118879 Chr.17: 19382320-19382320 on Build GRCh38
rs771770181 Chr.17: 19382385-19382385 on Build GRCh38
rs34992596 Chr.17: 19382399-19382399 on Build GRCh38
rs775109792 Chr.17: 19382207-19382207 on Build GRCh38
rs761624549 Chr.17: 19382220-19382220 on Build GRCh38
rs762526231 Chr.17: 19382286-19382286 on Build GRCh38
rs201095735 Chr.17: 19382195-19382195 on Build GRCh38
rs779993745 Chr.17: 19382236-19382236 on Build GRCh38
rs370236486 Chr.17: 19382181-19382181 on Build GRCh38
rs868484052 Chr.17: 19382273-19382273 on Build GRCh38
rs774843228 Chr.17: 19382185-19382185 on Build GRCh38
rs748486443 Chr.17: 19382248-19382248 on Build GRCh38
rs374009042 Chr.17: 19382374-19382374 on Build GRCh38
rs779283737 Chr.17: 19382242-19382242 on Build GRCh38

View Full Product Details