Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91145855-91146118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4 NM_133503.3 NM_133504.3 NM_133505.3 NM_133506.3 NM_133507.3 XM_005268693.1 XM_006719270.1 XM_017018917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256741 c.1048C>T p.R350C Chr.12: 91146090-91146090 on Build GRCh38
4045357 c.1049G>A p.R350H Chr.12: 91146089-91146089 on Build GRCh38
4456984 c.1029C>T p.T343T Chr.12: 91146109-91146109 on Build GRCh38
1364830 c.1045G>A p.V349M Chr.12: 91146093-91146093 on Build GRCh38
6226705 c.1044C>T p.Y348Y Chr.12: 91146094-91146094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7441 Chr.12: 91145898-91145898 on Build GRCh38
rs1042068978 Chr.12: 91145991-91145991 on Build GRCh38
rs770199273 Chr.12: 91145961-91145961 on Build GRCh38
rs767455312 Chr.12: 91146067-91146067 on Build GRCh38
rs1034558596 Chr.12: 91145901-91145901 on Build GRCh38
rs749620072 Chr.12: 91146032-91146032 on Build GRCh38
rs759810024 Chr.12: 91146053-91146053 on Build GRCh38
rs758251041 Chr.12: 91146088-91146088 on Build GRCh38
rs1014352244 Chr.12: 91145913-91145913 on Build GRCh38
rs1025628500 Chr.12: 91145955-91145955 on Build GRCh38
rs780518731 Chr.12: 91146095-91146095 on Build GRCh38
rs200191955 Chr.12: 91146093-91146093 on Build GRCh38
rs945036916 Chr.12: 91145982-91145982 on Build GRCh38
rs777182296 Chr.12: 91145899-91145899 on Build GRCh38
rs770306684 Chr.12: 91146021-91146021 on Build GRCh38
rs766286927 Chr.12: 91146089-91146089 on Build GRCh38
rs531541570 Chr.12: 91145978-91145978 on Build GRCh38
rs771344865 Chr.12: 91146039-91146039 on Build GRCh38
rs564091657 Chr.12: 91145910-91145910 on Build GRCh38
rs191434778 Chr.12: 91145953-91145953 on Build GRCh38
rs748563399 Chr.12: 91146008-91146008 on Build GRCh38
rs750454825 Chr.12: 91146084-91146084 on Build GRCh38
rs372887327 Chr.12: 91146055-91146055 on Build GRCh38
rs1803343 Chr.12: 91146007-91146007 on Build GRCh38
rs775666283 Chr.12: 91146072-91146072 on Build GRCh38
rs138760512 Chr.12: 91146073-91146073 on Build GRCh38
rs774379850 Chr.12: 91146040-91146040 on Build GRCh38
rs571211118 Chr.12: 91146004-91146004 on Build GRCh38
rs773757050 Chr.12: 91146027-91146027 on Build GRCh38
rs751102416 Chr.12: 91146090-91146090 on Build GRCh38
rs765239185 Chr.12: 91146076-91146076 on Build GRCh38

View Full Product Details