Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89112451-89112724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP2
Entrez Gene ID
2634
Gene Name
guanylate binding protein 2
Gene Aliases
-
UniGene
Hs.386567
Gene Chromosome Location
Chr.1: 89106132-89126159 on Build GRCh38
Transcripts
NM_004120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5649059 c.1318G>C p.E440Q Chr.1: 89112516-89112516 on Build GRCh38
682797 c.1207G>C p.D403H Chr.1: 89112627-89112627 on Build GRCh38
5759696 c.1341G>T p.Q447H Chr.1: 89112493-89112493 on Build GRCh38
5485970 c.1168C>T p.R390* Chr.1: 89112666-89112666 on Build GRCh38
5601540 c.1252G>A p.E418K Chr.1: 89112582-89112582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761974512 Chr.1: 89112557-89112557 on Build GRCh38
rs144298347 Chr.1: 89112572-89112572 on Build GRCh38
rs746735174 Chr.1: 89112631-89112631 on Build GRCh38
rs765107402 Chr.1: 89112578-89112580 on Build GRCh38
rs778592823 Chr.1: 89112528-89112528 on Build GRCh38
rs965939005 Chr.1: 89112632-89112632 on Build GRCh38
rs754371385 Chr.1: 89112593-89112593 on Build GRCh38
rs958982414 Chr.1: 89112579-89112579 on Build GRCh38
rs767575808 Chr.1: 89112561-89112561 on Build GRCh38
rs756561528 Chr.1: 89112478-89112478 on Build GRCh38
rs758553820 Chr.1: 89112640-89112640 on Build GRCh38
rs750660211 Chr.1: 89112580-89112580 on Build GRCh38
rs752094742 Chr.1: 89112602-89112602 on Build GRCh38
rs781006190 Chr.1: 89112640-89112640 on Build GRCh38
rs761023283 Chr.1: 89112683-89112683 on Build GRCh38
rs1018830576 Chr.1: 89112619-89112619 on Build GRCh38
rs373422153 Chr.1: 89112678-89112678 on Build GRCh38
rs768691309 Chr.1: 89112547-89112547 on Build GRCh38
rs567894250 Chr.1: 89112540-89112540 on Build GRCh38
rs777289203 Chr.1: 89112610-89112610 on Build GRCh38
rs747729921 Chr.1: 89112529-89112529 on Build GRCh38
rs139664617 Chr.1: 89112539-89112539 on Build GRCh38
rs1047606024 Chr.1: 89112519-89112519 on Build GRCh38
rs369240458 Chr.1: 89112653-89112653 on Build GRCh38
rs750332417 Chr.1: 89112628-89112630 on Build GRCh38
rs767822323 Chr.1: 89112472-89112472 on Build GRCh38
rs771598627 Chr.1: 89112535-89112535 on Build GRCh38
rs1013130657 Chr.1: 89112582-89112582 on Build GRCh38
rs372558261 Chr.1: 89112696-89112696 on Build GRCh38
rs746717769 Chr.1: 89112537-89112537 on Build GRCh38
rs761731691 Chr.1: 89112667-89112667 on Build GRCh38
rs757933226 Chr.1: 89112609-89112609 on Build GRCh38
rs761114907 Chr.1: 89112589-89112589 on Build GRCh38
rs774255077 Chr.1: 89112551-89112551 on Build GRCh38
rs193093983 Chr.1: 89112693-89112693 on Build GRCh38
rs914901197 Chr.1: 89112661-89112661 on Build GRCh38
rs754483732 Chr.1: 89112526-89112526 on Build GRCh38
rs770692588 Chr.1: 89112635-89112635 on Build GRCh38
rs773290360 Chr.1: 89112681-89112681 on Build GRCh38
rs779501679 Chr.1: 89112596-89112596 on Build GRCh38
rs754404117 Chr.1: 89112493-89112493 on Build GRCh38
rs1028819178 Chr.1: 89112668-89112668 on Build GRCh38
rs759519440 Chr.1: 89112543-89112543 on Build GRCh38
rs766904458 Chr.1: 89112592-89112592 on Build GRCh38
rs775509201 Chr.1: 89112665-89112665 on Build GRCh38
rs149847868 Chr.1: 89112536-89112536 on Build GRCh38
rs200030074 Chr.1: 89112594-89112594 on Build GRCh38
rs780704348 Chr.1: 89112490-89112490 on Build GRCh38
rs750732216 Chr.1: 89112476-89112476 on Build GRCh38
rs769669389 Chr.1: 89112657-89112657 on Build GRCh38
rs923714030 Chr.1: 89112612-89112612 on Build GRCh38

View Full Product Details