Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47836585-47836850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC46
Entrez Gene ID
90506
Gene Name
leucine rich repeat containing 46
Gene Aliases
-
UniGene
Hs.130767
Gene Chromosome Location
Chr.17: 47831627-47837713 on Build GRCh38
Transcripts
NM_033413.3 XM_005257775.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1684511 c.613_624del12 p.E205_L208delEQEL Chr.17: 47836767-47836778 on Build GRCh38
980539 c.635G>T p.R212M Chr.17: 47836789-47836789 on Build GRCh38
4443305 c.667C>A p.H223N Chr.17: 47836821-47836821 on Build GRCh38
3989090 c.687G>A p.M229I Chr.17: 47836841-47836841 on Build GRCh38
4556206 c.682G>A p.E228K Chr.17: 47836836-47836836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755834038 Chr.17: 47836738-47836738 on Build GRCh38
rs778558236 Chr.17: 47836778-47836778 on Build GRCh38
rs746201821 Chr.17: 47836797-47836797 on Build GRCh38
rs750176996 Chr.17: 47836737-47836737 on Build GRCh38
rs369363985 Chr.17: 47836740-47836740 on Build GRCh38
rs767273416 Chr.17: 47836730-47836730 on Build GRCh38
rs750092821 Chr.17: 47836712-47836715 on Build GRCh38
rs560767901 Chr.17: 47836631-47836631 on Build GRCh38
rs936894315 Chr.17: 47836639-47836639 on Build GRCh38
rs377191761 Chr.17: 47836767-47836778 on Build GRCh38
rs924103914 Chr.17: 47836637-47836637 on Build GRCh38
rs373351041 Chr.17: 47836798-47836798 on Build GRCh38
rs774195453 Chr.17: 47836716-47836716 on Build GRCh38
rs775737579 Chr.17: 47836802-47836802 on Build GRCh38
rs774476796 Chr.17: 47836826-47836826 on Build GRCh38
rs541109054 Chr.17: 47836607-47836607 on Build GRCh38
rs149736761 Chr.17: 47836651-47836651 on Build GRCh38
rs754588113 Chr.17: 47836767-47836767 on Build GRCh38
rs757722583 Chr.17: 47836795-47836795 on Build GRCh38
rs142960476 Chr.17: 47836759-47836770 on Build GRCh38
rs761806881 Chr.17: 47836725-47836725 on Build GRCh38
rs376083521 Chr.17: 47836634-47836634 on Build GRCh38
rs781411266 Chr.17: 47836796-47836796 on Build GRCh38
rs754945820 Chr.17: 47836698-47836698 on Build GRCh38
rs768791497 Chr.17: 47836808-47836808 on Build GRCh38
rs532247311 Chr.17: 47836755-47836755 on Build GRCh38
rs758123399 Chr.17: 47836745-47836745 on Build GRCh38
rs955824240 Chr.17: 47836793-47836793 on Build GRCh38
rs971073628 Chr.17: 47836736-47836736 on Build GRCh38
rs549324396 Chr.17: 47836658-47836658 on Build GRCh38
rs750494487 Chr.17: 47836659-47836659 on Build GRCh38
rs148647571 Chr.17: 47836807-47836807 on Build GRCh38
rs952276369 Chr.17: 47836649-47836649 on Build GRCh38
rs1027833569 Chr.17: 47836622-47836622 on Build GRCh38
rs747523264 Chr.17: 47836780-47836780 on Build GRCh38

View Full Product Details