Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130695443-130695716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5225486 c.123-1delG p.? Chr.9: 130695491-130695491 on Build GRCh38
7099076 c.185G>C p.R62T Chr.9: 130695554-130695554 on Build GRCh38
292948 c.123G>T p.R41R Chr.9: 130695492-130695492 on Build GRCh38
6657293 c.156C>T p.L52L Chr.9: 130695525-130695525 on Build GRCh38
4468892 c.156C>G p.L52L Chr.9: 130695525-130695525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988370294 Chr.9: 130695552-130695552 on Build GRCh38
rs1027565324 Chr.9: 130695572-130695572 on Build GRCh38
rs996451470 Chr.9: 130695533-130695533 on Build GRCh38
rs570396756 Chr.9: 130695472-130695472 on Build GRCh38
rs973816391 Chr.9: 130695476-130695476 on Build GRCh38
rs186124210 Chr.9: 130695480-130695480 on Build GRCh38
rs757697510 Chr.9: 130695613-130695613 on Build GRCh38
rs777097865 Chr.9: 130695522-130695522 on Build GRCh38
rs760555341 Chr.9: 130695609-130695609 on Build GRCh38
rs373286412 Chr.9: 130695555-130695555 on Build GRCh38
rs370483071 Chr.9: 130695596-130695596 on Build GRCh38
rs373224770 Chr.9: 130695469-130695469 on Build GRCh38
rs148157757 Chr.9: 130695531-130695531 on Build GRCh38
rs1043773952 Chr.9: 130695635-130695635 on Build GRCh38
rs574835336 Chr.9: 130695668-130695668 on Build GRCh38
rs550383127 Chr.9: 130695504-130695504 on Build GRCh38
rs1004106463 Chr.9: 130695679-130695679 on Build GRCh38
rs778394466 Chr.9: 130695505-130695505 on Build GRCh38
rs753509235 Chr.9: 130695611-130695611 on Build GRCh38
rs1023382128 Chr.9: 130695669-130695669 on Build GRCh38
rs755239894 Chr.9: 130695640-130695640 on Build GRCh38
rs760566353 Chr.9: 130695657-130695658 on Build GRCh38
rs375867059 Chr.9: 130695610-130695610 on Build GRCh38
rs776570598 Chr.9: 130695508-130695508 on Build GRCh38
rs887720575 Chr.9: 130695581-130695581 on Build GRCh38
rs377234336 Chr.9: 130695571-130695571 on Build GRCh38
rs113275970 Chr.9: 130695643-130695643 on Build GRCh38
rs754110717 Chr.9: 130695479-130695479 on Build GRCh38
rs199841784 Chr.9: 130695564-130695564 on Build GRCh38
rs765363231 Chr.9: 130695641-130695641 on Build GRCh38
rs1050898076 Chr.9: 130695654-130695654 on Build GRCh38
rs868325685 Chr.9: 130695674-130695674 on Build GRCh38
rs752164925 Chr.9: 130695490-130695490 on Build GRCh38
rs748977410 Chr.9: 130695554-130695554 on Build GRCh38
rs960373167 Chr.9: 130695662-130695662 on Build GRCh38
rs116465881 Chr.9: 130695665-130695665 on Build GRCh38
rs771446451 Chr.9: 130695521-130695521 on Build GRCh38
rs370282291 Chr.9: 130695560-130695560 on Build GRCh38
rs369974111 Chr.9: 130695497-130695497 on Build GRCh38
rs748117919 Chr.9: 130695503-130695503 on Build GRCh38
rs755257901 Chr.9: 130695649-130695649 on Build GRCh38
rs939587418 Chr.9: 130695541-130695541 on Build GRCh38
rs1015936712 Chr.9: 130695687-130695687 on Build GRCh38

View Full Product Details